J&W HP-PLOT U Columns, Agilent Technologies
An excellent column for C1 to C7 hydrocarbons, CO₂, methane, air/CO, water, oxygenates, amines, solvents, alcohols, ketones, and aldehydes.
- Improved resolution in less time than conventional packed columns
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W HP-PLOT U consists of bonded, divinylbenzene/ethylene glycol dimethacrylate coated onto a fused silica capillary. It is suitable for analyzing hydrocarbons (natural gas, refinery gas, C1 to C7, all C1–C3 isomers except propylene and propane), CO₂, methane, air/CO, water, and polar compounds.
This column features greater polarity than HP-PLOT Q (RI ethyl acetate 630 vs. 576), for different selectivity, better peak shape for some polar compounds such as water, and a much lower maximum operating temperature.
- Catalog No:
- CAAG19095P-UO4
- CAAG19091PUO4L
- CAAG19095PUO4E
- CAAG19091P-UO4
- CAAG19095P-UO3
- CAAG19095PUO4P
- USP code:
- G45
- G45
- G45
Frequently Bought Together