PL Rapide Aqua, Agilent Technologies
Analytique, 3 µm, 7.50 mm, 150 mm, Fully Porous
Catalog # CAAGPL11203830
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Supplier #: PL1120-3830
PL Rapide Aqua, Agilent Technologies
Catalog # CAAGPL11203830
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
The PL Rapide Aqua SEC columns offer high speed and resolution on older systems.
- PL Rapide Aqua SEC columns for high speed and resolution on older systems
- High-efficiency PL aquagel-OH media combined with wide-diameter hardware
- Increase overall efficiency by increasing flow rate and reducing run time
The PL Rapide Aqua SEC columns are configured to offer high speed and resolution on older systems by combining high-efficiency PL aquagel-OH media with wide-diameter hardware. By increasing flow rate and reducing run time, overall resolution is improved on systems with high dead volumes and large flow cell detectors.
Specifications
- Type de colonne:Analytique
- Ø int.:7.50 mm
- Granulométrie:3 µm
- Longueur:150 mm
- Packing Material:Fully Porous
- Cat. No.:CAAGPL11203830
- Max. pressure:150 bar
- Temperature limit:90 °C
- pH range:2 - 10
- Wetted materials:0.02% sodium azide