J&W Intuvo Low-bleed Columns, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
Designed for chromatography of a broad range of low-level analytes (for use with the Intuvo 9000 GC System).
- Unique combination of polysiloxane chemistry and proprietary surface deactivation
- General purpose columns with strict testing requirements for low bleed, inertness, selectivity, and efficiency
- Optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
- For use with the Agilent Intuvo 9000 GC System
Agilent J&W Low-bleed Intuvo GC/MS columns (compatible only with the Agilent Intuvo 9000 GC System) are designed for chromatography of a broad range of low-level analytes, and generate lower bleed even at high temperatures. The unique polymer chemistry and proprietary surface deactivation of these columns contribute to their adherence to the tightest quality control specifications in the industry for bleed, inertness, selectivity, and efficiency. Agilent J&W Low-bleed Intuvo GC/MS columns utilize special surface deactivation and siloxane chemistries that enhance the chromatographic performance of siloxane polymers.
Spécifications
- Catalog No:
- CAAGCP9205-INT
- CAAGCP9105-INT
- CAAGCP8944-INT
- CAAGCP9100-INT
- CAAGCP8982-INT
- Polarity:
- High-polarity stationary phase
- Mid-polarity stationary phase
- Low-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase