ContactUsLinkComponent
Sentinel XL® HP Half Hood Conversion Kit - CBRN to XL HP White Half Hood with 2 HEPA Filters, ILC Dover
Catalog # 76533-548
Supplier:  ILC DOVER LP
undefined
Sentinel XL® HP Half Hood Conversion Kit - CBRN to XL HP White Half Hood with 2 HEPA Filters, ILC Dover
Catalog # 76533-548
Supplier:  ILC DOVER LP
Supplier Number:  S-3000-05
Specifications
  • Description:
    Sentinel XL® HP Half hood conversion kit
  • Cat. no.:
    76533-548
Specifications
About this item
Convert the CBRN PAPR system into a HP PAPR system in a few easy steps.
  • Go from an APF of 25 to a NIOSH CAP2 protection rating in just minutes
  • Save time and space, just one blower needed for CBRN and HP settings
  • Easily change out hoods and filters
  • Latex-free
The blower stays the same, only the hood and filters must be changed. Kit is supplied with 33" breathing tube, half hood, plug, and 2 HEPA cartridge filters.