Masterflex® Reducer, Female Threaded Adapter, Straight, Avantor®
Fournisseur: MIDWEST VALVE & FITTING INC
Certificats
À propos de cet article
Maintain a permanent connection.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Sturdy threaded adapters are designed for permanent installation. Choose the size and material that fits your application needs.
- Environmentally Preferable:
Spécifications
- Model:
- 31410-13
- 31410-12
- 31211-10
- 31410-15
- 31410-14
- 31410-16
- Fitting depth:
- 8.73 mm
- 11.1125 mm
- 15.08 mm
- 11.1125 mm
- 11.1125 mm
- Sterilisation:
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year