Masterflex® I/P® Pump Head Adapters, Avantor® | Avantor
Masterflex® I/P® Pump Head Adapters, Avantor®
Masterflex® I/P® Pump Head Adapters, Avantor®
Certificates
Masterflex® I/P® Pump Head Adapters, Avantor®
Masterflex® I/P® Pump Head Adapters, Avantor®
4 Options Available Below
Product Details & Documents
Connect any I/P® pump head to a 56C frame motor.
  • Cast aluminum adapters feature built-in gear box for easy setup
  • Compatible with all I/P pump heads
  • Suitable for use with 1⁄10 hp to 1⁄2 hp 56C frame motor and 650 rpm max pump head speed
These I/P® pump head adapters work with 56C frame motors, allowing you to customize a pump for your application. Add a motor controller for variable-speed and/or remote-control operation. Adapters are compatible with all I/P pump heads.
Delivery information: Includes adapter with built-in gear box, hardware.
Specifications for Product Family
Spécifications
  • Model:
  • 77490-00
  • 77490-10
  • 77490-20
  • 77490-30
  • Drive height:
  • 165 mm (6.5")
  • 165 mm (6.5")
  • 165 mm (6.5")
  • 165 mm (6.5")
  • Drive length:
  • 152 mm (6")
  • 152 mm (6")
  • 171 mm (6.75")
  • 171 mm (6.75")
  • Drive width:
  • 165 mm (6.5")
  • 165 mm (6.5")
  • 165 mm (6.5")
  • 165 mm (6.5")
  • Gear ratio:
  • 3.1:1
  • 4.8:1
  • 9.7:1
  • 17.8:1
  • Min. RPM:
  • 20 rpm
  • 20 rpm
  • 20 rpm
  • 20 rpm
  • Motor mount:
  • NEMA Type 56C
  • NEMA Type 56C
  • NEMA Type 56C
  • NEMA Type 56C
  • Motor requirements:
  • 1/4 hp to 1/2 hp; 2015 max motor rpm
  • 1/4 hp to 1/2 hp; 3120 max motor rpm
  • 1/4 hp to 1/2 hp; 3600 max motor rpm
  • 1/4 hp to 1/2 hp; 4800 max motor rpm
  • Mounting type:
  • Direct coupling
  • Direct coupling
  • Direct coupling
  • Direct coupling
  • Open-head sensor:
  • No
  • No
  • No
  • No
  • Pump series:
  • I/P
  • I/P
  • I/P
  • I/P
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Débit maximal
    No. of Pump Heads Accepted
    Matériau du boîtier
    Max. RPM
    Availability
    Price
  • MFLX77490-00
    I/P NEMA Type 56 C Pump Head Adapter, 650 rpm max, 3.1:1 gear ratio
    17000 ml/min
    2
    Painted aluminium
    650 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,840.00
    $1,840.00
    Each
       $1,840.00
    Supplier Number: 77490-00
    Spécifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    MFLX77490-00
    Model
    77490-00
    Drive height
    165 mm (6.5")
    Drive length
    152 mm (6")
    Drive width
    165 mm (6.5")
    Gear ratio
    3.1:1
    Min. RPM
    20 rpm
    Motor mount
    NEMA Type 56C
    Motor requirements
    1/4 hp to 1/2 hp; 2015 max motor rpm
    Mounting type
    Direct coupling
    Open-head sensor
    No
    Pump series
    I/P
    Warranty
    2 years
    2 year warranty
  • MFLX77490-10
    I/P NEMA Type 56 C Pump Head Adapter, 650 rpm max, 4.8:1 gear ratio
    17000 ml/min
    2
    Painted aluminium
    650 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,840.00
    $1,840.00
    Each
       $1,840.00
    Supplier Number: 77490-10
    Spécifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    MFLX77490-10
    Model
    77490-10
    Drive height
    165 mm (6.5")
    Drive length
    152 mm (6")
    Drive width
    165 mm (6.5")
    Gear ratio
    4.8:1
    Min. RPM
    20 rpm
    Motor mount
    NEMA Type 56C
    Motor requirements
    1/4 hp to 1/2 hp; 3120 max motor rpm
    Mounting type
    Direct coupling
    Open-head sensor
    No
    Pump series
    I/P
    Warranty
    2 years
    2 year warranty
  • MFLX77490-20
    I/P NEMA Type 56 C Pump Head Adapter, 650 rpm max, 9.7:1 gear ratio
    9300 ml/min
    2
    Painted aluminium
    356 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,840.00
    $1,840.00
    Each
       $1,840.00
    Supplier Number: 77490-20
    Spécifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    MFLX77490-20
    Model
    77490-20
    Drive height
    165 mm (6.5")
    Drive length
    171 mm (6.75")
    Drive width
    165 mm (6.5")
    Gear ratio
    9.7:1
    Min. RPM
    20 rpm
    Motor mount
    NEMA Type 56C
    Motor requirements
    1/4 hp to 1/2 hp; 3600 max motor rpm
    Mounting type
    Direct coupling
    Open-head sensor
    No
    Pump series
    I/P
    Warranty
    2 years
    2 year warranty
  • MFLX77490-30
    I/P NEMA Type 56 C pump head adapter, 17,8:1 adapter gear ratio
    5000 ml/min
    2
    Painted aluminium
    194 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,840.00
    $1,840.00
    Each
       $1,840.00
    Supplier Number: 77490-30
    Spécifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    MFLX77490-30
    Model
    77490-30
    Drive height
    165 mm (6.5")
    Drive length
    171 mm (6.75")
    Drive width
    165 mm (6.5")
    Gear ratio
    17.8:1
    Min. RPM
    20 rpm
    Motor mount
    NEMA Type 56C
    Motor requirements
    1/4 hp to 1/2 hp; 4800 max motor rpm
    Mounting type
    Direct coupling
    Open-head sensor
    No
    Pump series
    I/P
    Warranty
    2 years
    2 year warranty