Masterflex® I/P® Variable-Speed Air-Powered Drive, Avantor®
I/P variable-speed air-powered drive., 17000 ml/min, 2, Painted steel, 650 rpm
Catalog # MFLX07589-30
CE Compliant
Supplier: Avantor Fluid Handling
Fournisseur #: 7589-30
Masterflex® I/P® Variable-Speed Air-Powered Drive, Avantor®
Catalog # MFLX07589-30
Supplier: Avantor Fluid Handling
CE Compliant
Jump to:
Product Details & Documents
Pump safely in hazardous
environments.
- Flow rate 0.6 to 13 LPM I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
- Easily adjust motor speed by using included regulator
- Effortlessly connect to your existing air supply or compressor
- The 3/4-hp air-powered motor starts smooth, runs cool, and requires little maintenance
- ATEX Zone 2 approved; rated to EEXII 3 G c IIC T6; NEC Class 1, Division 2, Groups A, B, C, D, T6
Use this variable-speed air-powered drive when electrical power may be unsafe or impractical. For safe operation, carefully ground to protect from static electricity.
Spécifications
- Débit maximal:17000 ml/min
- Désignation:I/P variable-speed air-powered drive.
- Max. RPM:650 rpm
- No. of Pump Heads Accepted:2
- Matériau du boîtier:Painted steel
- Open-head sensor:No
- Speed:Variable
- Drive length:282 mm (11.125")
- Motor hp:3/4
- Warranty:1 year
- Motor:560 W
- Reversible motor:No
- Type of pump heads accepted:Standard, Easy-Load, High-Performance
- Control type:Analog Variable Speed
- Drive width:216 mm (8.5")
- Cat. No.:MFLX07589-30
- Drive IP rating:IP 34
- Drive height:229 mm (9")
- Pump series:I/P
- Speed control:±10%
- Min. RPM:100 rpm