Masterflex® Multi-Syringe Pumps, Multi-Step, Avantor®
À propos de cet article
Ideal for low-flow, low-pressure applications where precision is needed.
- Flow ranges from 0.05 uL/min up to 70.56 ml/min
- Built-in program lets you enter up to eight commands to perform a variety of functions—you can even repeat commands up to ten times; all settings are stored in nonvolatile memory
- Backlit alphanumeric LCD ste
Program flow rate hold, pause, and change of direction; each command can be set to run for up to 12 hours. You can even program infusion/withdrawal pumps to sequence operations: either withdrawal followed by infusion or infusion followed by withdrawal. Ch
Delivery information: Includes a 6-ft cord with grounded plug (U.S. standard plug on 115 VAC model and European plug on 230 VAC models).
1 Options Available Below
Product Details & Documents
Ideal for low-flow, low-pressure applications where precision is needed.
- Flow ranges from 0.05 uL/min up to 70.56 ml/min
- Built-in program lets you enter up to eight commands to perform a variety of functions—you can even repeat commands up to ten times; all settings are stored in nonvolatile memory
- Backlit alphanumeric LCD ste
Program flow rate hold, pause, and change of direction; each command can be set to run for up to 12 hours. You can even program infusion/withdrawal pumps to sequence operations: either withdrawal followed by infusion or infusion followed by withdrawal. Ch
Delivery information: Includes a 6-ft cord with grounded plug (U.S. standard plug on 115 VAC model and European plug on 230 VAC models).
Specifications for Product Family
Spécifications
- Catalog No:
- DUP-DO-NOT-USE 490
- DUP-DO-NOT-USE 491
- MFLX74900-65
- DUP-DO-NOT-USE 080
- DUP-DO-NOT-USE 085
- MFLX74900-85
- DUP-DO-NOT-USE 010
- MFLX74901-05
- MFLX74901-15
- Model:
- 74900-60
- 74900-65
- 74900-65
- 74900-80
- 74900-85
- 74900-85
- 74901-00
- 74901-05
- 74901-15
- Accuracy:
- ±1.0%
- ±1.0%
- ±1.0%
- ±1.0%
- ±1.0%
- ±1.0%
- ±0.5%
- ±0.5%
- ±0.5%
- Input:
- Baud rate: four settings up to 9600
- Baud rate: four settings up to 9600
- Baud rate: four settings up to 9600
- Linear force:
- 40 lbs.
- 40 lbs.
- 40 lbs.
- 40 lbs.
- 40 lbs.
- 40 lbs.
- 40 lbs.
- 40 lbs.
- 40 lbs.
- Reproducibility:
- ±0.1%
- ±0.1%
- ±0.1%
- ±0.1%
- ±0.1%
- ±0.1%
- ±0.2%
- ±0.2%
- ±0.2%
- Syringe size accepted:
- 10 µl - 140 ml
- 10 µl - 140 ml
- 10 µl - 140 ml
- 10 µl - 140 ml
- 10 µl - 140 ml
- 10 µl - 140 ml
- 10 µl - 140 ml
- 10 µl - 140 ml
- 10 µl - 140 ml
- Syringe travel rate:
- <126.7"
- <126.7"
- <126.7"
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Spécifications
Product Family Options
Informations sur le produit
- DésignationHauteurLongueurWidthPuissanceMax. Flow Rate Per SyringeNo. of SyringesAvailabilityPrice
- Infusion Pump, Dual-Syringe, Programmable; 115 VAC15.2 cm (6")27.94 cm (11")228.6 mm (9")115 VAC2Discontinued-Spécifications:
More
Specifications
Cat. No.DUP-DO-NOT-USE 490Model74900-60Accuracy±1.0%InputLinear force40 lbs.Reproducibility±0.1%Syringe size accepted10 µl - 140 mlSyringe travel rateWarranty1 year - Infusion Pump, Dual-Syringe, Programmable; 230 VAC15.2 cm (6")27.94 cm (11")228.6 mm (9")230 VAC2Discontinued-Spécifications:CE Compliant
More
Specifications
Cat. No.DUP-DO-NOT-USE 491Model74900-65Accuracy±1.0%InputLinear force40 lbs.Reproducibility±0.1%Syringe size accepted10 µl - 140 mlSyringe travel rateWarranty1 yearCE Compliant - Infusion Pump, Dual-Syringe, Programmable; 230 VAC15.2 cm (6")27.94 cm (11")228.6 mm (9")230 VAC2Log in pour la disponibilitéEach$7,980.00Spécifications:CE CompliantIrregular Voltage
More
Specifications
Cat. No.MFLX74900-65Model74900-65Accuracy±1.0%InputLinear force40 lbs.Reproducibility±0.1%Syringe size accepted10 µl - 140 mlSyringe travel rateWarranty1 yearCE CompliantIrregular Voltage - Infusion Pump, Ten-Syringe Capacity; 115 VAC 50/60 Hz15.2 cm (6")27.94 cm (11")304.8 mm (12")115 VAC10Discontinued-Spécifications:
More
Specifications
Cat. No.DUP-DO-NOT-USE 080Model74900-80Accuracy±1.0%InputLinear force40 lbs.Reproducibility±0.1%Syringe size accepted10 µl - 140 mlSyringe travel rateWarranty1 year - Infusion Pump, Ten-Syringe Capacity; 230 VAC 50/60 Hz15.2 cm (6")27.94 cm (11")304.8 mm (12")230 VAC10Discontinued-Spécifications:CE Compliant
More
Specifications
Cat. No.DUP-DO-NOT-USE 085Model74900-85Accuracy±1.0%InputLinear force40 lbs.Reproducibility±0.1%Syringe size accepted10 µl - 140 mlSyringe travel rateWarranty1 yearCE Compliant - Infusion Pump, Ten-Syringe Capacity; 230 VAC 50/60 Hz15.2 cm (6")27.94 cm (11")304.8 mm (12")230 VAC10Discontinued-Spécifications:CE CompliantIrregular Voltage
More
Specifications
Cat. No.MFLX74900-85Model74900-85Accuracy±1.0%InputLinear force40 lbs.Reproducibility±0.1%Syringe size accepted10 µl - 140 mlSyringe travel rateWarranty1 yearCE CompliantIrregular Voltage - Infusion/Withdrawal Pump, Dual-Syringe, Programmable; 115 VAC23.5 cm (9.25")27.94 cm (11")127 mm (5")115 VAC147 ml/min2Discontinued-Spécifications:
More
Specifications
Cat. No.DUP-DO-NOT-USE 010Model74901-00Accuracy±0.5%InputBaud rate: four settings up to 9600Linear force40 lbs.Reproducibility±0.2%Syringe size accepted10 µl - 140 mlSyringe travel rate<126.7"Warranty1 year - Infusion/Withdrawal pump, Dual-Syringe, Programmable; 230 VAC23.5 cm (9.25")27.94 cm (11")127 mm (5")230 VAC147 ml/min2Discontinued-Spécifications:CE CompliantIrregular Voltage
More
Specifications
Cat. No.MFLX74901-05Model74901-05Accuracy±0.5%InputBaud rate: four settings up to 9600Linear force40 lbs.Reproducibility±0.2%Syringe size accepted10 µl - 140 mlSyringe travel rate<126.7"WarrantyCE CompliantIrregular Voltage - Infusion/Withdrawal Pump, Ten-Syringe, Programmable; 230 VAC30.48 cm (12")27.94 cm (11")127 mm (5")230 VAC147 ml/min10Discontinued-Spécifications:CE Compliant
More
Specifications
Cat. No.MFLX74901-15Model74901-15Accuracy±0.5%InputBaud rate: four settings up to 9600Linear force40 lbs.Reproducibility±0.2%Syringe size accepted10 µl - 140 mlSyringe travel rate<126.7"Warranty1 yearCE Compliant
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Pumps, Syringe Pumps Daisy Chain Cable for 74901-80 Dual Drive Syringe PumpPumps, Syringe Pumps Daisy Chain Cable for 74901-80 Dual Drive Syringe PumpMFLX74901-87Log in pour la disponibilitéEach - $130.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Pumps, Syringe Pumps RS-232 Connector Cable for 74901-80 Dual Drive Syringe PumpPumps, Syringe Pumps RS-232 Connector Cable for 74901-80 Dual Drive Syringe PumpMFLX74901-88Log in pour la disponibilitéEach - $321.00