Masterflex® L/S® Precision Variable-Speed Modular Pump Systems, Avantor®
About this item
Place where convenient—ideal for limited spaces, hoods, or isolation chambers.
- Flow rate 0.06 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm and tubing size
- Three digit LED display shows motor rpm — confirm speed setting at a glance
- Reversible motor—purge tubing before or after pumping; pump fluid in either direction
- LED indicators confirm motor direction
- Choose between benchtop controller or washdown wall-mount controller drives
L/S® modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
Benchtop controller drives feature a splash-resistant ABS plastic controller housing with IP33 rating and remote control capabilities via a DB9 female connector on back of controller. Wall-mount controller housing is powder-coated aluminum and IP66/NEMA 4X rated for washdown with remote control capabilities via weather-resistant connection on controller.
3 Options Available Below
Product Details & Documents
Place where convenient—ideal for limited spaces, hoods, or isolation chambers.
- Flow rate 0.06 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm and tubing size
- Three digit LED display shows motor rpm — confirm speed setting at a glance
- Reversible motor—purge tubing before or after pumping; pump fluid in either direction
- LED indicators confirm motor direction
- Choose between benchtop controller or washdown wall-mount controller drives
L/S® modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
Benchtop controller drives feature a splash-resistant ABS plastic controller housing with IP33 rating and remote control capabilities via a DB9 female connector on back of controller. Wall-mount controller housing is powder-coated aluminum and IP66/NEMA 4X rated for washdown with remote control capabilities via weather-resistant connection on controller.
Specifications for Product Family
Specifications
- Catalog No:
- MFLX77913-60
- MFLX77913-70
- MFLX77913-80
- MFLX77913-60-AU
- MFLX77913-60-CH
- MFLX77913-60-DK
- MFLX77913-60-EU
- MFLX77913-60-IL
- MFLX77913-60-IN
- MFLX77913-60-IT
- MFLX77913-60-UK
- MFLX77913-70-AU
- MFLX77913-70-CH
- MFLX77913-70-DK
- MFLX77913-70-EU
- MFLX77913-70-IL
- MFLX77913-70-IN
- MFLX77913-70-IT
- MFLX77913-70-UK
- MFLX77913-80-AU
- MFLX77913-80-CH
- MFLX77913-80-DK
- MFLX77913-80-EU
- MFLX77913-80-IL
- MFLX77913-80-IN
- MFLX77913-80-IT
- MFLX77913-80-UK
- Model:
- 77913-60
- 77913-70
- 77913-80
- 77913-60
- 77913-60
- 77913-60
- 77913-60
- 77913-60
- 77913-60
- 77913-60
- 77913-60
- 77913-70
- 77913-70
- 77913-70
- 77913-70
- 77913-70
- 77913-70
- 77913-70
- 77913-70
- 77913-80
- 77913-80
- 77913-80
- 77913-80
- 77913-80
- 77913-80
- 77913-80
- 77913-80
- No. of channels:
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- No. of rollers:
- 3
- 4
- 4
- 3
- 3
- 3
- 3
- 3
- 3
- 3
- 3
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- 4
- Control type:
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Variable-Speed Digital
- Min. flow rate per channel:
- 4.8 ml/min
- 17 ml/min
- 17 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- Max. flow rate per channel:
- 480 ml/min
- 1700 ml/min
- 1700 ml/min
- 480 ml/min
- 480 ml/min
- 480 ml/min
- 480 ml/min
- 480 ml/min
- 480 ml/min
- 480 ml/min
- 480 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- 1700 ml/min
- Min. flow rate:
- 4.8 ml/min
- 17 ml/min
- 17 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 4.8 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- 17 ml/min
- Drive IP rating:
- IP 33
- IP 33
- IP 66
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 33
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- Accuracy:
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- ±0.25%
- Anti-drip:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Cloud connected:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Digital dispensing:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Drive:
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)
- 07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)
- 07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)
- 07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)
- 07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)
- 07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)
- 07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)
- 07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)
- 07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)
- Duty cycle:
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Min. RPM:
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- 6 rpm
- Open-head sensor:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Pump series:
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- Remote I/O:
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- 4 to 20 mA, 0 to 10 V, start/stop and direction via contact closure
- Reversible motor:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Tubing:
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
- Tubing sizes accepted:
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- L/S® 15, 24, 35, 36
- Washdown capable:
- No
- No
- Yes
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Current:
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- Frequency:
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- Voltage:
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
Specifications
Product Family Options
US plug models
Product Information
- DésignationPlug TypeDébit maximalHauteurLongueurWidthPump HeadPuissanceMax. RPMMatériau du boîtierAvailabilityPrice
- L/S Precision Modular Pump w/ Easy-Load 3 Head; 600 rpmUS480 ml/min114.3 mm (4.5")351.3 mm (13.83")111.3 mm (4.38")77800-60 (L/S® Easy-Load® 3 pump head)90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V600 rpmPainted steelLog in for availabilityEach$4,160.00Supplier Number: 77913-60Specifications:
More
Specifications
Cat. No.MFLX77913-60Model77913-60No. of channels1No. of rollers3Control typeVariable-Speed DigitalMin. flow rate per channel4.8 ml/minMax. flow rate per channel480 ml/minMin. flow rate4.8 ml/minDrive IP ratingIP 33Accuracy±0.25%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPump seriesL/SRemote I/O4 to 20 mA. 0 to 10 V. start/stop and direction via contact closureReversible motorYesTubing06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableNoCurrent2.2 A @ 115 V; 1.1 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 years - L/S Precision Modular Pump w/ Easy-Load II Head; 600 rpmUS1700 ml/min114.3 mm (4.5")338.6 mm (13.33")111.3 mm (4.38")77200-62 (L/S® Easy-Load® II pump head)90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V600 rpmPainted steelLog in for availabilityEach$4,270.00Supplier Number: 77913-70Specifications:
More
Specifications
Cat. No.MFLX77913-70Model77913-70No. of channels1No. of rollers4Control typeVariable-Speed DigitalMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 33Accuracy±0.25%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07557-00 (L/S® Digital Precision Modular Drive with Remote I/O and Benchtop Controller)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPump seriesL/SRemote I/O4 to 20 mA. 0 to 10 V. start/stop and direction via contact closureReversible motorYesTubing06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableNoCurrent2.2 A @ 115 V; 1.1 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 years - L/S Washdown Modular Pump w/ Easy-Load II Head; 600 rpmUS1700 ml/min152.4 mm (6")460.8 mm (18.14")177.8 mm (7")77200-62 (L/S® Easy-Load® II pump head)90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V600 rpmPainted steel and aluminiumLog in for availabilityEach$6,330.00Supplier Number: 77913-80Specifications:2 year warrantyCE CompliantIP66 washdown
More
Specifications
Cat. No.MFLX77913-80Model77913-80No. of channels1No. of rollers4Control typeVariable-Speed DigitalMin. flow rate per channel17 ml/minMax. flow rate per channel1700 ml/minMin. flow rate17 ml/minDrive IP ratingIP 66Accuracy±0.25%Anti-dripNoCloud connectedNoDigital dispensingNoDrive07557-60 (L/S® Digital Precision Washdown Modular Drive with Remote I/O and Wall-Mount Controller)Duty cycleContinuousMin. RPM6 rpmOpen-head sensorNoPump seriesL/SRemote I/O4 to 20 mA. 0 to 10 V. start/stop and direction via contact closureReversible motorYesTubing06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)Tubing sizes acceptedL/S® 15, 24, 35, 36Washdown capableYesCurrent2.2 A @ 115 V; 1.1 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 years2 year warrantyCE CompliantIP66 washdown