Masterflex® L/S® Digital Process Pump Systems, Avantor® | Avantor
Masterflex® L/S® Digital Process Pump Systems, Avantor®
Masterflex® L/S® Digital Process Pump Systems, Avantor®
Certificates
Masterflex® L/S® Digital Process Pump Systems, Avantor®
Masterflex® L/S® Digital Process Pump Systems, Avantor®
2 Options Available Below
Product Details & Documents
Transfer, dispense, and meter in lab, process and washdown environments.
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Models with open-head sensor stops the drive when the pump head is opened (when used with sensor-equipped pump head)
  • NEMA 4X (IP66) rated housing for washdown applications
  • Models with open-head sensor stop drive motor when pump head is opened
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
Specifications for Product Family
Spécifications
  • Model:
  • 77975-25
  • 77975-27
  • 77975-29
  • 77975-45
  • 77975-47
  • 77975-49
  • 77975-25
  • 77975-25
  • 77975-25
  • 77975-25
  • 77975-25
  • 77975-25
  • 77975-25
  • 77975-25
  • 77975-27
  • 77975-27
  • 77975-27
  • 77975-27
  • 77975-27
  • 77975-27
  • 77975-27
  • 77975-27
  • 77975-29
  • 77975-29
  • 77975-29
  • 77975-29
  • 77975-29
  • 77975-29
  • 77975-29
  • 77975-29
  • 77975-45
  • 77975-45
  • 77975-45
  • 77975-45
  • 77975-45
  • 77975-45
  • 77975-45
  • 77975-45
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • No. of rollers:
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • 4
  • Control type:
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Digital
  • Min. flow rate per channel:
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.08 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • Max. flow rate per channel:
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min (1.7 L/min)
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 480 ml/min
  • 480 ml/min
  • 480 ml/min
  • 480 ml/min
  • 480 ml/min
  • 480 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • 1700 ml/min
  • Min. flow rate:
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.08 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • 0.28 ml/min
  • Drive IP rating:
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • IP 66
  • Accuracy:
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • ±0.1% (Resolution: 0.1 rpm at 600 rpm)
  • Anti-drip:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Cloud connected:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Digital dispensing:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Drive:
  • 07575-30 (L/S® Digital Process Drive)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-40 (L/S® Digital Process Drive)
  • 07575-48 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-48 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-30 (L/S® Digital Process Drive)
  • 07575-30 (L/S® Digital Process Drive)
  • 07575-30 (L/S® Digital Process Drive)
  • 07575-30 (L/S® Digital Process Drive)
  • 07575-30 (L/S® Digital Process Drive)
  • 07575-30 (L/S® Digital Process Drive)
  • 07575-30 (L/S® Digital Process Drive)
  • 07575-30 (L/S® Digital Process Drive)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
  • 07575-40 (L/S® Digital Process Drive)
  • 07575-40 (L/S® Digital Process Drive)
  • 07575-40 (L/S® Digital Process Drive)
  • 07575-40 (L/S® Digital Process Drive)
  • 07575-40 (L/S® Digital Process Drive)
  • 07575-40 (L/S® Digital Process Drive)
  • 07575-40 (L/S® Digital Process Drive)
  • 07575-40 (L/S® Digital Process Drive)
  • Duty cycle:
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Min. RPM:
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • 0.1 rpm
  • Open-head sensor:
  • No
  • Yes
  • Yes
  • No
  • Yes
  • Yes
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • Remote I/O:
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • Reversible motor:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Tubing:
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • Tubing sizes accepted:
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • Washdown capable:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Current:
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • Frequency:
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • Voltage:
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Spécifications
Product Family Options
US plug models
Informations sur le produit
  • Désignation
    Plug Type
    Débit maximal
    Hauteur
    Longueur
    Width
    Pump Head
    Puissance
    Max. RPM
    Matériau du boîtier
    Availability
    Price
  • MFLX77975-25
    L/S® Digital Process Drive with Easy-Load® II Pump Head for High-Performance Tubing, Stainless Steel Housing; 90 to 260 VAC
    US
    1700 ml/min
    236.5 mm (9.31")
    370.3 mm (14.58")
    228.6 mm (9")
    77200-62 (L/S® Easy-Load® II pump head)
    90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V
    600 rpm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10,700.00
    $10,700.00
    Each
       $10,700.00
    Supplier Number: 77975-25
    Spécifications:
    2 year warranty
    CE Compliant
    IP66 washdown
    More
    Specifications
    Cat. No.
    MFLX77975-25
    Model
    77975-25
    No. of channels
    1
    No. of rollers
    4
    Control type
    Digital
    Min. flow rate per channel
    0.28 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    0.28 ml/min
    Drive IP rating
    IP 66
    Accuracy
    ±0.1% (Resolution: 0.1 rpm at 600 rpm)
    Anti-drip
    Yes
    Cloud connected
    No
    Digital dispensing
    Yes
    Drive
    07575-30 (L/S® Digital Process Drive)
    Duty cycle
    Continuous
    Min. RPM
    0.1 rpm
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
    Reversible motor
    Yes
    Tubing
    06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    Yes
    Current
    2.2 A @ 115 V; 1.1 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
    IP66 washdown
  • MFLX77975-27
    L/S® Digital Process Drive with Open-Head Sensor and Easy-Load® II Pump Head for Precision Tubing, Stainless Steel Housing; 90 to 260 VAC
    US
    480 ml/min
    77200-30 (L/S® Easy-Load® II head with open-head sensor)
    90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V
    600 rpm
    Stainless steel
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX77975-27
    Model
    77975-27
    No. of channels
    1
    No. of rollers
    4
    Control type
    Digital
    Min. flow rate per channel
    0.28 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    0.28 ml/min
    Drive IP rating
    IP 66
    Accuracy
    ±0.1% (Resolution: 0.1 rpm at 600 rpm)
    Anti-drip
    Yes
    Cloud connected
    No
    Digital dispensing
    Yes
    Drive
    07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
    Duty cycle
    Continuous
    Min. RPM
    0.1 rpm
    Open-head sensor
    Yes
    Pump series
    L/S
    Remote I/O
    0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
    Reversible motor
    Yes
    Tubing
    06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    Yes
    Current
    2.2 A @ 115 V; 1.1 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
  • MFLX77975-29
    L/S® Digital Process Drive with Open-Head Sensor and Easy-Load® II Pump Head for High-Performance Tubing, Stainless Steel Housing; 90 to 260 VAC
    US
    1700 ml/min
    77200-32 (L/S® Easy-Load® II head with open-head sensor)
    90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V
    600 rpm
    Stainless steel
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX77975-29
    Model
    77975-29
    No. of channels
    1
    No. of rollers
    4
    Control type
    Digital
    Min. flow rate per channel
    0.28 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    0.28 ml/min
    Drive IP rating
    IP 66
    Accuracy
    ±0.1% (Resolution: 0.1 rpm at 600 rpm)
    Anti-drip
    Yes
    Cloud connected
    No
    Digital dispensing
    Yes
    Drive
    07575-38 (L/S® Digital Process Drive with Open-Head Sensor)
    Duty cycle
    Continuous
    Min. RPM
    0.1 rpm
    Open-head sensor
    Yes
    Pump series
    L/S
    Remote I/O
    0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
    Reversible motor
    Yes
    Tubing
    06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    Yes
    Current
    2.2 A @ 115 V; 1.1 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
  • MFLX77975-45
    L/S® Digital Process Drive with Easy-Load® II Pump Head for High-Performance Tubing, Powder-Coated Steel Housing; 90 to 260 VAC
    US
    1700 ml/min
    236.5 mm (9.31")
    370.3 mm (14.58")
    228.6 mm (9")
    77200-62 (L/S® Easy-Load® II pump head)
    90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V
    600 rpm
    Powder-coated steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,660.00
    $9,660.00
    Each
       $9,660.00
    Supplier Number: 77975-45
    Spécifications:
    2 year warranty
    CE Compliant
    IP66 washdown
    More
    Specifications
    Cat. No.
    MFLX77975-45
    Model
    77975-45
    No. of channels
    1
    No. of rollers
    4
    Control type
    Digital
    Min. flow rate per channel
    0.28 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    0.28 ml/min
    Drive IP rating
    IP 66
    Accuracy
    ±0.1% (Resolution: 0.1 rpm at 600 rpm)
    Anti-drip
    Yes
    Cloud connected
    No
    Digital dispensing
    Yes
    Drive
    07575-40 (L/S® Digital Process Drive)
    Duty cycle
    Continuous
    Min. RPM
    0.1 rpm
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
    Reversible motor
    Yes
    Tubing
    06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    Yes
    Current
    2.2 A @ 115 V; 1.1 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
    IP66 washdown
  • MFLX77975-47
    L/S® Digital Process Drive with Open-Head Sensor and Easy-Load® II Pump Head for Precision Tubing, Powder-Coated Steel Housing; 90 to 260 VAC
    US
    480 ml/min
    77200-30 (L/S® Easy-Load® II head with open-head sensor)
    90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V
    600 rpm
    Powder-coated steel
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX77975-47
    Model
    77975-47
    No. of channels
    1
    No. of rollers
    4
    Control type
    Digital
    Min. flow rate per channel
    0.08 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    0.08 ml/min
    Drive IP rating
    IP 66
    Accuracy
    ±0.1% (Resolution: 0.1 rpm at 600 rpm)
    Anti-drip
    Yes
    Cloud connected
    No
    Digital dispensing
    Yes
    Drive
    07575-48 (L/S® Digital Process Drive with Open-Head Sensor)
    Duty cycle
    Continuous
    Min. RPM
    0.1 rpm
    Open-head sensor
    Yes
    Pump series
    L/S
    Remote I/O
    0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Reversible motor
    Yes
    Tubing
    06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    Yes
    Current
    2.2 A @ 115 V; 1.1 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
  • MFLX77975-49
    L/S® Digital Process Drive with Open-Head Sensor and Easy-Load® II Pump Head for High-Performance Tubing, Powder-Coated Steel Housing; 90 to 260 VAC
    US
    1700 ml/min
    77200-32 (L/S® Easy-Load® II head with open-head sensor)
    90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V
    600 rpm
    Powder-coated steel
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX77975-49
    Model
    77975-49
    No. of channels
    1
    No. of rollers
    4
    Control type
    Digital
    Min. flow rate per channel
    0.28 ml/min
    Max. flow rate per channel
    1700 ml/min (1.7 L/min)
    Min. flow rate
    0.28 ml/min
    Drive IP rating
    IP 66
    Accuracy
    ±0.1% (Resolution: 0.1 rpm at 600 rpm)
    Anti-drip
    Yes
    Cloud connected
    No
    Digital dispensing
    Yes
    Drive
    07575-48 (L/S® Digital Process Drive with Open-Head Sensor)
    Duty cycle
    Continuous
    Min. RPM
    0.1 rpm
    Open-head sensor
    Yes
    Pump series
    L/S
    Remote I/O
    0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Reversible motor
    Yes
    Tubing
    06434-24, 10 ft (C-FLEX ULTRA®, L/S® 24 )
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    Yes
    Current
    2.2 A @ 115 V; 1.1 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years