Masterflex® L/S® Precision Pump Tubing, C-Flex®, Avantor®
Masterflex® L/S® Precision Pump Tubing, C-Flex®, Avantor®
Masterflex® L/S® Precision Pump Tubing, C-Flex®, Avantor®
Masterflex® L/S® Precision Pump Tubing, C-Flex®, Avantor®
Combines biocompatibility of silicone with chemical resistance similar to Tygon®.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Very low protein binding
  • Heat sealable, weldable
  • Economical tubing
Applications: General-purpose lab uses requiring high performance and long life
Characteristics: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, opaque white
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
  • Model:
  • 06424-13
  • 06424-14
  • 06424-16
  • 06424-17
  • 06424-18
  • 06424-25
  • Length per pack:
  • 7.6 m (25 ft.)
  • 7.6 m (25 ft.)
  • 7.6 m (25 ft.)
  • 7.6 m (25 ft.)
  • 7.6 m (25 ft.)
  • 7.6 m (25 ft.)
  • Max. pressure:
  • 2.7 bar (40 psi)
  • 2.7 bar (40 psi)
  • 2.7 bar (40 psi)
  • 1.4 bar (20 psi)
  • 0.7 bar (10 psi)
  • 1.4 bar (20 psi)
  • Min. temperature:
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • Max. temperature:
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • Min. temperature (dynamic):
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Max. temperature (dynamic):
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Max. vacuum:
  • 660 mm Hg (26" Hg)
  • 660 mm Hg (26" Hg)
  • 660 mm Hg (26" Hg)
  • 510 mm Hg (20" Hg)
  • 510 mm Hg (20" Hg)
  • 660 mm Hg (26" Hg)
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • Regulatory approvals:
  • ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
  • ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
  • ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
  • ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
  • ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
  • ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
  • Sterilisation:
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Sterilisation method autoclave:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method EtO:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method gamma:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Suction lift H₂O:
  • 8.8 m (29 ft.)
  • 8.8 m (29 ft.)
  • 8.8 m (29 ft.)
  • 6.7 m (22 ft.)
  • 6.7 m (22 ft.)
  • 8.8 m (29 ft.)
Product Family Options
  • Désignation
    Tubing Material
    Tube Ø int.
    Hose Barb Size
    L/S Masterflex Tubing Size
    Débit maximal
  • L/S® Precision Pump Tubing, C-Flex®, L/S 13; 25 ft
    C-Flex®
    0.80 mm (0.031")
    0.0625"
    13
    36 ml/min
    CE certificate
    Specifications
    Cat. No.
    MFLX06424-13
    Model
    06424-13
    Length per pack
    7.6 m (25 ft.)
    Max. pressure
    2.7 bar (40 psi)
    Min. temperature
    –45 °C (–50 °F)
    Max. temperature
    121 °C (250 °F)
    Min. temperature (dynamic)
    –40 °C (–40 °F)
    Max. temperature (dynamic)
    100 °C (212 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide, Gamma Irradiation
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    Yes
    Suction lift H₂O
    8.8 m (29 ft.)
    CE certificate
  • L/S® Precision Pump Tubing, C-Flex®, L/S 14; 25 ft
    C-Flex®
    1.60 mm (0.063")
    0.0625"
    14
    130 ml/min
    Each (25Feet)
    Each (25Feet) - $98.90
    $98.90
    Each (25Feet)
       $98.90
    CE certificate
    Specifications
    Cat. No.
    MFLX06424-14
    Model
    06424-14
    Length per pack
    7.6 m (25 ft.)
    Max. pressure
    2.7 bar (40 psi)
    Min. temperature
    –45 °C (–50 °F)
    Max. temperature
    121 °C (250 °F)
    Min. temperature (dynamic)
    –40 °C (–40 °F)
    Max. temperature (dynamic)
    100 °C (212 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide, Gamma Irradiation
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    Yes
    Suction lift H₂O
    8.8 m (29 ft.)
    CE certificate
  • L/S® Precision Pump Tubing, C-Flex®, L/S 16; 25 ft
    C-Flex®
    3.10 mm (0.122")
    0.125"
    16
    480 ml/min
    Each (25Feet)
    Each (25Feet) - $106.00
    $106.00
    Each (25Feet)
       $106.00
    CE certificate
    Specifications
    Cat. No.
    MFLX06424-16
    Model
    06424-16
    Length per pack
    7.6 m (25 ft.)
    Max. pressure
    2.7 bar (40 psi)
    Min. temperature
    –45 °C (–50 °F)
    Max. temperature
    121 °C (250 °F)
    Min. temperature (dynamic)
    –40 °C (–40 °F)
    Max. temperature (dynamic)
    100 °C (212 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide, Gamma Irradiation
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    Yes
    Suction lift H₂O
    8.8 m (29 ft.)
    CE certificate
  • L/S® Precision Pump Tubing, C-Flex®, L/S 17; 25 ft
    C-Flex®
    6.40 mm (0.252")
    0.25"
    17
    1700 ml/min
    Each (25Feet)
    Each (25Feet) - $145.00
    $145.00
    Each (25Feet)
       $145.00
    CE certificate
    Specifications
    Cat. No.
    MFLX06424-17
    Model
    06424-17
    Length per pack
    7.6 m (25 ft.)
    Max. pressure
    1.4 bar (20 psi)
    Min. temperature
    –45 °C (–50 °F)
    Max. temperature
    121 °C (250 °F)
    Min. temperature (dynamic)
    –40 °C (–40 °F)
    Max. temperature (dynamic)
    100 °C (212 °F)
    Max. vacuum
    510 mm Hg (20" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide, Gamma Irradiation
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    Yes
    Suction lift H₂O
    6.7 m (22 ft.)
    CE certificate
  • L/S® Precision Pump Tubing, C-Flex®, L/S 18; 25 ft
    C-Flex®
    7.90 mm (0.311")
    0.375"
    18
    2300 ml/min
    Each (25Feet)
    Each (25Feet) - $160.00
    $160.00
    Each (25Feet)
       $160.00
    CE certificate
    Specifications
    Cat. No.
    MFLX06424-18
    Model
    06424-18
    Length per pack
    7.6 m (25 ft.)
    Max. pressure
    0.7 bar (10 psi)
    Min. temperature
    –45 °C (–50 °F)
    Max. temperature
    121 °C (250 °F)
    Min. temperature (dynamic)
    –40 °C (–40 °F)
    Max. temperature (dynamic)
    100 °C (212 °F)
    Max. vacuum
    510 mm Hg (20" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide, Gamma Irradiation
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    Yes
    Suction lift H₂O
    6.7 m (22 ft.)
    CE certificate
  • L/S® Precision Pump Tubing, C-Flex®, L/S 25; 25 ft
    C-Flex®
    4.80 mm (0.189")
    0.1875"
    25
    1000 ml/min
    Each (25Feet)
    Each (25Feet) - $137.00
    $137.00
    Each (25Feet)
       $137.00
    CE certificate
    Specifications
    Cat. No.
    MFLX06424-25
    Model
    06424-25
    Length per pack
    7.6 m (25 ft.)
    Max. pressure
    1.4 bar (20 psi)
    Min. temperature
    –45 °C (–50 °F)
    Max. temperature
    121 °C (250 °F)
    Min. temperature (dynamic)
    –40 °C (–40 °F)
    Max. temperature (dynamic)
    100 °C (212 °F)
    Max. vacuum
    660 mm Hg (26" Hg)
    Pump series
    L/S
    Regulatory approvals
    ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
    Sterilisation
    Autoclave, Ethylene Oxide, Gamma Irradiation
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    Yes
    Suction lift H₂O
    8.8 m (29 ft.)
    CE certificate