Masterflex® High-Flow Submersible Centrifugal Pumps, Avantor®
Pump liquids with suspended solids up to 2" diameter at flow rates up to 175 GPM.
- Stainless steel pumps are chemical and corrosion resistant
- Manually-operated pumps allow for continuous emptying and automatically-operated pumps are used in on-demand dewatering applications
- Dual-seal design ensures rugged and reliable service
Submersible pumps are ideal for food and beverage processing; irrigation and storm water runoff; spraying; and water and wastewater treatment applications. Operate pumps either fully or partially submerged.
- Environmentally Preferable:
- Model:
- 75505-50
- 75505-55
- 75505-57
- 75505-70
- 75505-75
- Duty cycle continuous:
- Yes
- Yes
- Yes
- Yes
- Yes
- Inlet connection:
- No screen
- No screen
- No screen
- No screen
- No screen
- Max. temperature:
- 40 °C (104 °F)
- 40 °C (104 °F)
- 40 °C (104 °F)
- 40 °C (104 °F)
- 40 °C (104 °F)
- Motor type:
- Washdown
- Washdown
- Washdown
- Washdown
- Washdown
- Motor hp:
- 1/2
- 1/2
- 1/2
- 1
- 1
- Outlet connection:
- 2 NPT(F)
- 2 NPT(F)
- 2 NPT(F)
- 2 NPT(F)
- 2 NPT(F)
- Wetted materials:
- 304 SS pump head impeller, shaft, and housing; carbon-ceramic and silicon-carbide seals; and Viton O-rings
- 304 SS pump head impeller, shaft, and housing; carbon-ceramic and silicon-carbide seals; and Viton O-rings
- 304 SS pump head impeller, shaft, and housing; carbon-ceramic and silicon-carbide seals; and Viton O-rings
- 304 SS pump head impeller, shaft, and housing; carbon-ceramic and silicon-carbide seals; and Viton O-rings
- 304 SS pump head impeller, shaft, and housing; carbon-ceramic and silicon-carbide seals; and Viton O-rings
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Frequently Bought Together