Masterflex® L/S® Variable-Speed Economy Modular Drives, Avantor®
Masterflex® L/S® Variable-Speed Economy Modular Drives, Avantor®
Certificates
Masterflex® L/S® Variable-Speed Economy Modular Drives, Avantor®
Masterflex® L/S® Variable-Speed Economy Modular Drives, Avantor®
4 Options Available Below
Product Details & Documents
Versatile variable-speed drive with a wide flow range and simple controls.
  • Flow rate 0.06 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Simple operation with single-turn speed control
  • Separate motor and controller are connected via a 6-ft (1.9 m) cable — place components where convenient or where space allows
  • Single-turn speed control and on/off switch with green power indicator — maintain speed setting while turning pump on/off
  • The 1/10-hp reversible motor has soft start and back EMF for ±2% speed control
  • Painted steel housing is IP33 rated for protection from vertically falling water
A perfect upgrade from a fixed-speed drive, these variable-speed drives offer a wider rpm range, ±2% speed regulation, and ETL and CE listings. For added versatility, drives accept all L/S pump heads.
 : Includes 6-ft (1.8-m) line cord; 115 VAC: U.S. standard plug; 230 VAC: IEC 320/CEE22 socket—pumps are shipped with cord/plug set specific to destination country.
Specifications for Product Family
Spécifications
  • Model:
  • 07559-00
  • 07559-05
  • 07559-10
  • 07559-15
  • Cloud connected:
  • No
  • No
  • No
  • No
  • Controller height:
  • 87 mm (3.4")
  • 87 mm (3.4")
  • 87 mm (3.4")
  • 87 mm (3.4")
  • Controller IP rating:
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • Controller length:
  • 204 mm (8")
  • 204 mm (8")
  • 204 mm (8")
  • 204 mm (8")
  • Controller width:
  • 164 mm (6.4")
  • 164 mm (6.4")
  • 164 mm (6.4")
  • 164 mm (6.4")
  • Control type:
  • Analog Variable Speed
  • Analog Variable Speed
  • Analog Variable Speed
  • Analog Variable Speed
  • Digital dispensing:
  • No
  • No
  • No
  • No
  • Drive IP rating:
  • IP 33
  • IP 33
  • IP 33
  • Min. RPM:
  • 6 rpm
  • 6 rpm
  • 1 rpm
  • 1 rpm
  • Motor:
  • 75 W
  • 75 W
  • 75 W
  • 75 W
  • Motor height:
  • 115 mm (4.5")
  • 115 mm (4.5")
  • 115 mm (4.5")
  • 115 mm (4.5")
  • Motor hp:
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • Motor IP rating:
  • IP 21
  • IP 21
  • IP 21
  • IP 21
  • Motor length:
  • 198 mm (7.8")
  • 198 mm (7.8")
  • 198 mm (7.8")
  • 198 mm (7.8")
  • Motor width:
  • 97 mm (3.7968")
  • 97 mm (3.7968")
  • 97 mm (3.7968")
  • 97 mm (3.7968")
  • Open-head sensor:
  • No
  • No
  • No
  • No
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • Remote I/O:
  • None
  • None
  • None
  • None
  • Reversible motor:
  • Yes
  • Yes
  • Yes
  • Yes
  • Speed:
  • Variable
  • Variable
  • Variable
  • Variable
  • Speed control:
  • ±2%
  • ±2%
  • ±2%
  • ±2%
  • Type of pump heads accepted:
  • Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
  • Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
  • Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
  • Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
  • Washdown capable:
  • No
  • No
  • No
  • No
  • Current:
  • 1.2 A
  • 1.2 A
  • 2.2 A
  • 1.2 A
  • Frequency:
  • 60 Hz
  • 50 Hz
  • 60 Hz
  • 50 Hz
  • Voltage:
  • 115 VAC
  • 230 VAC
  • 115 VAC
  • 230 VAC
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Spécifications
Product Family Options
US plug models
Informations sur le produit
  • Désignation
    Plug Type
    Débit maximal
    No. of Pump Heads Accepted
    Matériau du boîtier
    Max. RPM
    Puissance
    Availability
    Price
  • MFLX07559-00
    L/S Variable-speed modular drive, 6 - 600 rpm, 115 VAC
    US
    3400 ml/min
    2
    Painted steel and aluminium
    600 rpm
    115 VAC; 60 Hz; 1.2 A
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,300.00
    $2,300.00
    Each
       $2,300.00
    Supplier Number: 7559-00
    Spécifications:
    2 year warranty
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX07559-00
    Model
    07559-00
    Cloud connected
    No
    Controller height
    87 mm (3.4")
    Controller IP rating
    IP 33
    Controller length
    204 mm (8")
    Controller width
    164 mm (6.4")
    Control type
    Analog Variable Speed
    Digital dispensing
    No
    Drive IP rating
    Min. RPM
    6 rpm
    Motor
    75 W
    Motor height
    115 mm (4.5")
    Motor hp
    1/10
    Motor IP rating
    IP 21
    Motor length
    198 mm (7.8")
    Motor width
    97 mm (3.7968")
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    None
    Reversible motor
    Yes
    Speed
    Variable
    Speed control
    ±2%
    Type of pump heads accepted
    Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
    Washdown capable
    No
    Current
    1.2 A
    Frequency
    60 Hz
    Voltage
    115 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
  • MFLX07559-05
    L/S Variable-speed modular drive, 6 - 600 rpm, 230 VAC
    US
    3400 ml/min
    2
    Painted steel and aluminium
    600 rpm
    230 VAC; 50 Hz; 1.2 A
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,340.00
    $2,340.00
    Each
       $2,340.00
    Supplier Number: 7559-05
    Spécifications:
    2 year warranty
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX07559-05
    Model
    07559-05
    Cloud connected
    No
    Controller height
    87 mm (3.4")
    Controller IP rating
    IP 33
    Controller length
    204 mm (8")
    Controller width
    164 mm (6.4")
    Control type
    Analog Variable Speed
    Digital dispensing
    No
    Drive IP rating
    IP 33
    Min. RPM
    6 rpm
    Motor
    75 W
    Motor height
    115 mm (4.5")
    Motor hp
    1/10
    Motor IP rating
    IP 21
    Motor length
    198 mm (7.8")
    Motor width
    97 mm (3.7968")
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    None
    Reversible motor
    Yes
    Speed
    Variable
    Speed control
    ±2%
    Type of pump heads accepted
    Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
    Washdown capable
    No
    Current
    1.2 A
    Frequency
    50 Hz
    Voltage
    230 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
  • MFLX07559-10
    L/S Variable-speed modular drive, 1 - 100 rpm, 115 VAC
    US
    580 ml/min
    4
    Painted steel and aluminium
    100 rpm
    115 VAC; 60 Hz; 2.2 A
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,300.00
    $2,300.00
    Each
       $2,300.00
    Supplier Number: 7559-10
    Spécifications:
    2 year warranty
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX07559-10
    Model
    07559-10
    Cloud connected
    No
    Controller height
    87 mm (3.4")
    Controller IP rating
    IP 33
    Controller length
    204 mm (8")
    Controller width
    164 mm (6.4")
    Control type
    Analog Variable Speed
    Digital dispensing
    No
    Drive IP rating
    IP 33
    Min. RPM
    1 rpm
    Motor
    75 W
    Motor height
    115 mm (4.5")
    Motor hp
    1/10
    Motor IP rating
    IP 21
    Motor length
    198 mm (7.8")
    Motor width
    97 mm (3.7968")
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    None
    Reversible motor
    Yes
    Speed
    Variable
    Speed control
    ±2%
    Type of pump heads accepted
    Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
    Washdown capable
    No
    Current
    2.2 A
    Frequency
    60 Hz
    Voltage
    115 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
  • MFLX07559-15
    L/S Variable-speed modular drive, 1 - 100 rpm, 230 VAC
    US
    580 ml/min
    4
    Painted steel and aluminium
    100 rpm
    230 VAC; 50 Hz; 1.2 A
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,340.00
    $2,340.00
    Each
       $2,340.00
    Supplier Number: 7559-15
    Spécifications:
    2 year warranty
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX07559-15
    Model
    07559-15
    Cloud connected
    No
    Controller height
    87 mm (3.4")
    Controller IP rating
    IP 33
    Controller length
    204 mm (8")
    Controller width
    164 mm (6.4")
    Control type
    Analog Variable Speed
    Digital dispensing
    No
    Drive IP rating
    IP 33
    Min. RPM
    1 rpm
    Motor
    75 W
    Motor height
    115 mm (4.5")
    Motor hp
    1/10
    Motor IP rating
    IP 21
    Motor length
    198 mm (7.8")
    Motor width
    97 mm (3.7968")
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    None
    Reversible motor
    Yes
    Speed
    Variable
    Speed control
    ±2%
    Type of pump heads accepted
    Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm
    Washdown capable
    No
    Current
    1.2 A
    Frequency
    50 Hz
    Voltage
    230 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant