Masterflex® L/S® MiniFlex® Pump Heads, Avantor®
Masterflex® L/S® MiniFlex® Pump Heads, Avantor®
Masterflex® L/S® MiniFlex® Pump Heads, Avantor®
Certificates
Masterflex® L/S® MiniFlex® Pump Heads, Avantor®
Masterflex® L/S® MiniFlex® Pump Heads, Avantor®
2 Options Available Below
Product Details & Documents
Pump head accepts continuous lengths of tubing for clean flow paths.
  • Flow range of 0.005 to 381 ml/min - flow rate depends on drive rpm, pump head, and tubing size
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Dual-channel model delivers synchronous flow from two separate channels; single-channel model delivers highest flow rate
  • Automatic tubing retention makes tube loading quick and easy
  • Suitable for use with Masterflex® L/S® Digital Miniflex® Pump Systems
The Masterflex® Miniflex® pump heads feature a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life. A compact overall size envelope makes these heads ideal for OEM applications and integration into fluid systems and machinery.
Specifications for Product Family
Specifications
  • Model:
  • 77220-20
  • 77220-30
  • Gear ratio:
  • 1:1
  • 1:1
  • Max. flow rate per channel:
  • 381 ml/min
  • 184 ml/min
  • Min. flow rate per channel:
  • 0.005 ml/min
  • 0.005 ml/min
  • No. of channels:
  • 1
  • 2
  • Occlusion:
  • Fixed
  • Fixed
  • Open-head sensor:
  • Yes
  • Yes
  • Optimum RPM range:
  • 0.1 - 300 rpm
  • 0.1 - 300 rpm
  • Pump series:
  • L/S
  • L/S
  • Tubing retention:
  • Automatic
  • Automatic
  • Tubing sizes accepted:
  • L/S® 13, L/S 14, L/S 16, L/S 25
  • L/S® 13, L/S 14, L/S 16
  • Volume per revolution with L/S 13 tubing:
  • 0.04 ml
  • 0.04 ml
  • Volume per revolution with L/S 14 tubing:
  • 0.14 ml
  • 0.14 ml
  • Volume per revolution with L/S 16 tubing:
  • 0.5 ml
  • 0.5 ml
  • Volume per revolution with L/S 25 tubing:
  • 1.1 ml
  • Warranty:
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • Désignation
    Rollers
    Matériau du boîtier
    Rotor Material
    Hauteur
    Longueur
    Width
    Availability
    Price
  • MFLX77220-20
    L/S Single-Channel Miniflex Pump Head, for 07525-series Pumps
    3 (thermoplastic polyester)
    Polyarylamide (PARA)
    Stainless steel and aluminum
    76 mm (3")
    70 mm (2.75")
    64 mm (2.5")
    Log in for availability
    Log in for availability
    Each
    Each - $426.00
    $426.00
    Each
       $426.00
    Supplier Number: 77220-20
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX77220-20
    Model
    77220-20
    Gear ratio
    1:1
    Max. flow rate per channel
    381 ml/min
    Min. flow rate per channel
    0.005 ml/min
    No. of channels
    1
    Occlusion
    Fixed
    Open-head sensor
    Yes
    Optimum RPM range
    0.1 - 300 rpm
    Pump series
    L/S
    Tubing retention
    Automatic
    Tubing sizes accepted
    L/S® 13, L/S 14, L/S 16, L/S 25
    Volume per revolution with L/S 13 tubing
    0.04 ml
    Volume per revolution with L/S 14 tubing
    0.14 ml
    Volume per revolution with L/S 16 tubing
    0.5 ml
    Volume per revolution with L/S 25 tubing
    1.1 ml
    Warranty
    1 year
  • MFLX77220-30
    L/S Dual-Channel Miniflex Pump Head, for 07525-series Pumps
    3 (thermoplastic polyester)
    Polyarylamide (PARA)
    Stainless steel and aluminum
    76 mm (3")
    70 mm (2.75")
    64 mm (2.5")
    Log in for availability
    Log in for availability
    Each
    Each - $457.00
    $457.00
    Each
       $457.00
    Supplier Number: 77220-30
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX77220-30
    Model
    77220-30
    Gear ratio
    1:1
    Max. flow rate per channel
    184 ml/min
    Min. flow rate per channel
    0.005 ml/min
    No. of channels
    2
    Occlusion
    Fixed
    Open-head sensor
    Yes
    Optimum RPM range
    0.1 - 300 rpm
    Pump series
    L/S
    Tubing retention
    Automatic
    Tubing sizes accepted
    L/S® 13, L/S 14, L/S 16
    Volume per revolution with L/S 13 tubing
    0.04 ml
    Volume per revolution with L/S 14 tubing
    0.14 ml
    Volume per revolution with L/S 16 tubing
    0.5 ml
    Volume per revolution with L/S 25 tubing
    Warranty