Masterflex® L/S® Digital Process Pump Systems with Cytoflow™ Pump Head, Avantor®
À propos de cet article
Ideal for the transfer and dispensing of live cell cultures, media, and other shear-sensitive fluids in process and washdown environments.
- Convex-roller design is gentler on live cells and shear-sensitive fluids—validated by independent test data
- Large-diameter rotor delivers higher flow rates at lower speeds
- Low-speed pumping lets you maintain the consistency and characteristics of shear-sensitive fluids
- Drive housings are completely sealed—IP66 and NEMA 4X rated—simply hose down to clean
- Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
- Cytoflow pump head offers the best option for pumping live cells and shear-sensitive fluids--validated by independent test data
- Full remote control capabilities allow easy integration into automated processes
- Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually
Developed specifically for pumping delicate live cells and shear-sensitive fluids, the Masterflex L/S Cytoflow pump head is ideal for use in biopharma and microbiology applications where peristaltic pump operators want to minimize cell hemolysis. The extra-large tubing bed combined with fewer rollers results in high flow rates at low motor speeds, which furthers its cell viability advantages.
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
2 Options Available Below
Product Details & Documents
Ideal for the transfer and dispensing of live cell cultures, media, and other shear-sensitive fluids in process and washdown environments.
- Convex-roller design is gentler on live cells and shear-sensitive fluids—validated by independent test data
- Large-diameter rotor delivers higher flow rates at lower speeds
- Low-speed pumping lets you maintain the consistency and characteristics of shear-sensitive fluids
- Drive housings are completely sealed—IP66 and NEMA 4X rated—simply hose down to clean
- Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
- Cytoflow pump head offers the best option for pumping live cells and shear-sensitive fluids--validated by independent test data
- Full remote control capabilities allow easy integration into automated processes
- Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually
Developed specifically for pumping delicate live cells and shear-sensitive fluids, the Masterflex L/S Cytoflow pump head is ideal for use in biopharma and microbiology applications where peristaltic pump operators want to minimize cell hemolysis. The extra-large tubing bed combined with fewer rollers results in high flow rates at low motor speeds, which furthers its cell viability advantages.
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
Specifications for Product Family
Spécifications
- Catalog No:
- MFLX77975-50
- MFLX77975-52
- MFLX77975-50-AU
- MFLX77975-50-CH
- MFLX77975-50-DK
- MFLX77975-50-EU
- MFLX77975-50-IL
- MFLX77975-50-IN
- MFLX77975-50-IT
- MFLX77975-50-UK
- MFLX77975-52-AU
- MFLX77975-52-CH
- MFLX77975-52-DK
- MFLX77975-52-EU
- MFLX77975-52-IL
- MFLX77975-52-IN
- MFLX77975-52-IT
- MFLX77975-52-UK
- Model:
- 77975-50
- 77975-52
- 77975-50
- 77975-50
- 77975-50
- 77975-50
- 77975-50
- 77975-50
- 77975-50
- 77975-50
- 77975-52
- 77975-52
- 77975-52
- 77975-52
- 77975-52
- 77975-52
- 77975-52
- 77975-52
- No. of channels:
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- No. of rollers:
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- Control type:
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Digital
- Min. flow rate per channel:
- 0.17 ml/min
- 0.62 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- Max. flow rate per channel:
- 1040 ml/min
- 3700 ml/min
- 1040 ml/min
- 1040 ml/min
- 1040 ml/min
- 1040 ml/min
- 1040 ml/min
- 1040 ml/min
- 1040 ml/min
- 1040 ml/min
- 3700 ml/min
- 3700 ml/min
- 3700 ml/min
- 3700 ml/min
- 3700 ml/min
- 3700 ml/min
- 3700 ml/min
- 3700 ml/min
- Min. flow rate:
- 0.17 ml/min
- 0.62 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.17 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- 0.62 ml/min
- Drive IP rating:
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- IP 66
- Accuracy:
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- ±0.1% (Resolution: 0.1 rpm at 600 rpm)
- Anti-drip:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Cloud connected:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Digital dispensing:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Drive:
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- 07575-30 (L/S® Digital Process Drive)
- Duty cycle:
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Min. RPM:
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- 0,1 rpm
- Open-head sensor:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Pump series:
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- Remote I/O:
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
- Reversible motor:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Tubing:
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- 06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)
- Tubing sizes accepted:
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- L/S® 13, 14, 16, 25, 17, 18
- Washdown capable:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Current:
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- 2.2 A @ 115 V; 1.1 A @ 230 V
- Frequency:
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- 50/60 Hz
- Voltage:
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- 90 - 260 VAC
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
Spécifications
Product Family Options
US plug models
Informations sur le produit
- DésignationPlug TypeDébit maximalHauteurLongueurWidthPump HeadPuissanceMax. RPMMatériau du boîtierAvailabilityPrice
- L/S® Digital Process Drive with Cytoflow™ Pump Head for Precision Tubing; 90 to 260 VACUS1040 ml/min236.5 mm (9.31")370.3 mm (14.58")228.6 mm (9")77253-00 (L/S® Cytoflow™ head)90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V600 rpmStainless steelLog in pour la disponibilitéEach11 810,00 $Supplier Number: 77975-50Spécifications:2 year warranty
More
Specifications
Cat. No.MFLX77975-50Model77975-50No. of channels1No. of rollers2Control typeDigitalMin. flow rate per channel0.17 ml/minMax. flow rate per channel1040 ml/minMin. flow rate0.17 ml/minDrive IP ratingIP 66Accuracy±0.1% (Resolution: 0.1 rpm at 600 rpm)Anti-dripYesCloud connectedNoDigital dispensingYesDrive07575-30 (L/S® Digital Process Drive)Duty cycleContinuousMin. RPM0,1 rpmOpen-head sensorNoPump seriesL/SRemote I/O0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closureReversible motorYesTubing06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableYesCurrent2.2 A @ 115 V; 1.1 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 years2 year warranty - L/S® Digital Process Drive with Cytoflow™ Pump Head for High-Performance Tubing; 90 to 260 VACUS3700 ml/min236.5 mm (9.31")370.3 mm (14.58")228.6 mm (9")77253-02 (L/S® Cytoflow™ head)90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V600 rpmStainless steelLog in pour la disponibilitéEach11 810,00 $Supplier Number: 77975-52Spécifications:2 year warranty
More
Specifications
Cat. No.MFLX77975-52Model77975-52No. of channels1No. of rollers2Control typeDigitalMin. flow rate per channel0.62 ml/minMax. flow rate per channel3700 ml/minMin. flow rate0.62 ml/minDrive IP ratingIP 66Accuracy±0.1% (Resolution: 0.1 rpm at 600 rpm)Anti-dripYesCloud connectedNoDigital dispensingYesDrive07575-30 (L/S® Digital Process Drive)Duty cycleContinuousMin. RPM0,1 rpmOpen-head sensorNoPump seriesL/SRemote I/O0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closureReversible motorYesTubing06434-16, 10-ft (C-FLEX® Ultra, L/S® 16)Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18Washdown capableYesCurrent2.2 A @ 115 V; 1.1 A @ 230 VFrequency50/60 HzVoltage90 - 260 VACWarranty2 years2 year warranty