Masterflex® Pump Handheld Remote Controllers, Avantor® | Avantor
Masterflex® Pump Handheld Remote Controllers, Avantor®
Masterflex® Pump Handheld Remote Controllers, Avantor®
Certificates
Masterflex® Pump Handheld Remote Controllers, Avantor®
Masterflex® Pump Handheld Remote Controllers, Avantor®
3 Options Available Below
Product Details & Documents
Easily control your Masterflex® L/S® pumps while filling and dispensing.
  • Suitable for use with Masterflex® L/S® pumps
  • Long 10ft cable makes remote control simple
These ergonomic handheld controllers are designed to control basic pump drive operations including start/stop, direction, and prime — depending on available drive functions. Route tubing through the handle for convenience in filling and dispensing applications. These handheld controllers accept Masterflex tubing sizes L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18, L/S 15, and L/S 24 (up to 7/16" nominal OD).
Delivery information: Includes 6-ft (1.8 m) cable with noted terminal connector.
Specifications for Product Family
Spécifications
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • MFLX07523-97
    Handheld Remote Controller; DB25(M) Connector; 6 ft (1,8 m) Cable
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $700.00
    $700.00
    Each
       $700.00
    Supplier Number: 7523-97
    Spécifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX07523-97
    Warranty
    1 year
    CE Compliant
  • MFLX07528-80
    Handheld Remote Controller; DB9(M) Connector; 6 ft (1,8 m) Cable
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $689.00
    $689.00
    Each
       $689.00
    Supplier Number: 7528-80
    Spécifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX07528-80
    Warranty
    1 year
    CE Compliant
  • MFLX07595-62
    Handheld Remote Controller; Stripped Wire Ends; 6 ft (1,8 m) Cable
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $686.00
    $686.00
    Each
       $686.00
    Supplier Number: 7595-62
    Spécifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX07595-62
    Warranty
    1 year
    CE Compliant