Micropump Pump Head Adapter Kits
Micropump Pump Head Adapter Kits
Supplier:   MICROPUMP INC
Micropump Pump Head Adapter Kits
Supplier:  MICROPUMP INC
Certificates
Micropump Pump Head Adapter Kits
3 Options Available Below
Product Details & Documents
Easily adapt your pump head to your drive with these adapter kits.
  • Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.
Specifications for Product Family
Specifications
  • Warranty:
  • 1 year
Specifications
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • MFLX07003-99
    Gear Pump Head Adapter Kit, 56C face drive, standard magnets
    Log in for availability
    Log in for availability
    Each
    Each - $870.00
    $870.00
    Each
       $870.00
    Supplier Number: 81518
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX07003-99
    Warranty
  • MFLX07104-48
    Series Q pump head to drive adapter kit
    Log in for availability
    Log in for availability
    Each
    Each - $1,330.00
    $1,330.00
    Each
       $1,330.00
    Supplier Number: QPM
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX07104-48
    Warranty
    1 year
  • MFLX07003-96
    Pump Head Adapter Kit; NEMA 56C to A-Mount, High-Torque Mag
    Log in for availability
    Log in for availability
    Each
    Each - $1,540.00
    $1,540.00
    Each
       $1,540.00
    Supplier Number: 81518+213003
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX07003-96
    Warranty