Masterflex® High-Viscosity Sanitary Drum Pumps, Avantor®
Supplier: VWR International
Certificates
About this item
Capable of pumping fluids with viscosities up to 10,000 cps.
- Progressive cavity design allows for low-shear properties
- Flow rates up to 12 GPM (45 LPM)
- 316 stainless steel construction with PTFE elastomers
These drum pumps are engineered specifically for transferring food products, pharmaceuticals, and cosmetic products from barrels, kettles, and tanks. They offer a progressive cavity, positive-displacement delivery system resulting in a continuous flow of material with little product degradation. This unique design allows you to quickly dismantle, clean, and re-assemble the pump.
Delivery: Includes drive and pump tube.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX75220-00
- MFLX75220-20
- MFLX75220-30
- Model:
- 75220-00
- 75220-20
- 75220-30
- 3A sanitary regulatory approval:
- Yes
- Yes
- Yes
- CE regulatory approval:
- Yes
- Yes
- Yes
- Max. viscosity:
- 10000 cP
- 10000 cP
- 10000 cP
- Motor type:
- TEFC
- TEFC
- Air
- Motor hp:
- 1
- 1-1/2
- Tube length, inlet:
- 39"
- 39"
- 39"
- UL Canada regulatory approval:
- Yes
- Yes
- Yes
- UL US regulatory approval:
- Yes
- Yes
- Yes
- Warranty:
- 3 years
- 1 year
- 1 year