Masterflex® Electro-Optic Point-Level Sensors, Avantor®
Supplier: VWR International
Certificates
About this item
No clog or wear—contain no moving parts.
- Detect liquids regardless of conductivity, color, or specific gravity
- Economical and compact — built-in solid-state switching electronics
- One-unit installation — simply screws into the top or side of your tank
These dependable, electro-optic level sensors contain no moving parts to clog or wear. Sensors offer ±1 mm repeatability and are available in a wide variety of materials to meet even the most demanding applications.
Installation is simple and quick through the tank top, bottom, or side. The switches can be used for low-, intermediate-, or high-liquid level detection. Solid state switching ensures dependability over a long service life. Order controller separately.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX32772-30
- MFLX32772-32
- MFLX32772-08
- MFLX32772-10
- MFLX32772-12
- MFLX32772-20
- MFLX32772-00
- MFLX32772-22
- MFLX32772-02
- MFLX32772-04
- Model:
- 32772-30
- 32772-32
- 32772-08
- 32772-10
- 32772-12
- 32772-20
- 32772-00
- 32772-22
- 32772-02
- 32772-04
- Max. process temperature:
- 100 °C
- 100 °C
- 80 °C
- 80 °C
- 100 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- Max. pressure:
- 2500 psi
- 2500 psi
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- Process mount:
- 1/2"NPT(M)
- 1/2"NPT(M)
- 1/2" Straight Thread
- 1/2" Straight Thread
- 3/8" NPT(M)
- Flanged
- 1/4" NPT(M)
- Flanged
- 1/4" NPT(M)
- 1/4" NPT(M)
- Relay output:
- 30 VDC at 40 mA max
- 30 VDC at 40 mA max
- 30 VDC at 40 mA max
- 30 VDC at 40 mA max
- 30 VDC at 40 mA max
- 30 VDC at 40 mA max
- 30 VDC at 40 mA max
- 30 VDC at 40 mA max
- 30 VDC at 40 mA max
- 30 VDC at 40 mA max
- Wetted materials:
- Nickel-plated Carbon steel/Fused glass
- Nickel-plated Carbon steel/Fused glass
- Polysulfone
- Polysulfone
- Isoplast
- Polysulfone
- Polysulfone
- Polysulfone
- Polysulfone
- Nylon
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year