Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems, Avantor® | Avantor
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems, Avantor®
Supplier:   VWR International
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems, Avantor®
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems, Avantor®
3 Options Available Below
Product Details & Documents
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Single-use pump heads are constructed with a disposable polypropylene chamber
  • Reusable pump heads are constructed of durable 316L stainless steel
Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.
Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.
Specifications for Product Family
Spécifications
  • Warranty:
  • Shelf life
  • 1 year
  • 1 year
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • MFLX76000-02
    Disposable Pump Chamber; PP machined for stainless steel single-use Quattroflow pump systems
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,940.00
    $2,940.00
    Each
       $2,940.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX76000-02
    Warranty
    Shelf life
  • MFLX76000-06
    Multiple-Use Pump Chamber; 316L machined for ABS and stainless steel multiuse Quattroflow pump systems
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,410.00
    $3,410.00
    Each
       $3,410.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX76000-06
    Warranty
    1 year
  • MFLX76000-08
    Mutiple-Use Pump Chamber; 316L machined stainless steel multiuse Quattroflow pump systems
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,500.00
    $6,500.00
    Each
       $6,500.00
    Spécifications:
    More
    Specifications
    Cat. No.
    MFLX76000-08
    Warranty
    1 year
Accessories
Informations sur le produit
  • Catalog #
    Plug Type
    Maximum Flow Rate
    Availability
    Price
  • Multi-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VAC
    Multi-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VAC
    MFLX74000-49
    US
    3 L/min
    Log in pour la disponibilité
    Each - $16,860.00