Masterflex® Adhesive-Coated Microscope Slides, Avantor®
Supplier:   VWR International
Masterflex® Adhesive-Coated Microscope Slides, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Adhesive-Coated Microscope Slides, Avantor®
Masterflex® Adhesive-Coated Microscope Slides, Avantor®
1 Options Available Below
Product Details & Documents
Don't let your samples slip away.
  • Adhesive coatings fix cells and tissues to slides for hybridization and other staining techniques
  • Extensive quality control methods ensure each batch performs optimally
  • Coating formulations are effective without interfering with staining
Use these adhesive-coated microscope slides when sample adhesion is critical for proper staining and reliable results.
A. Gelatin is the most common adhesive coating used in histology, ideal for simple stains.
B. Poly-l-lysine is a stickier coating for more advanced immunostaining, but may result in precipitation during staining by silver impregnation methods.
C. Silane offers superior adhesion, perfect for applications requiring antigen retrieval, and resists bleaching and enzymatic predigestion.
D. Hybridization slides feature an adhesive coating that withstands organic solvents and a variety of temperatures, best suited to in situ hybridization applications.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Sterile
    Surface Treatment
    L×l
    Availability
    Price
  • MFLX75955-45
    Adhesive coated microscope slides, Poly-l-lysine
    Glass
    -
    Poly-D-Lysine
    76.2×25.4 mm (3×1")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $248.00
    $248.00
    Log in pour la disponibilité
    Pack of 144
    Pack of 144 - $172.22
    $172.22
    Each
       $248.00
    Pack of 144
       $172.22
  • MFLX75955-47
    Adhesive coated microscope slides, Silane
    Glass
    -
    Silane
    76.2×25.4 mm (3×1")
    Discontinued
  • MFLX75955-49
    Adhesive coated microscope slides, hybridization
    Glass
    -
    Hybridization-specific
    76.2×25.4 mm (3×1")
    Discontinued
  • MFLX75955-51
    Adhesive coated microscope slides, gelatin
    Glass
    -
    Gelatin
    76.2×25.4 mm (3×1")
    Discontinued