VWR® Advanced Hotplates
Hotplate 7×7", 120 V, 10 L, Ceramic, square, Max. 500 °C
470330-728
These top-of-the-line units combine advanced digital features with high quality and reliability.
- All models feature digital displays
- Prominent 'Hot' indicator
- Rocker switch for added safety
- All models include a 5 year warranty
These high quality units feature advanced digital displays with a prominent hot surface indicator, a rocker switch do disconnect the heater element, audio alarm and overheat protection. Advanced electronics provide high temperature stability (±1%). The new design reduces benchtop footprint by over 20%, and all models come with a probe connection capability for even greater accuracy.
The temperature is adjustable in 1 °C increments, with equivalent readability. The enclosure is made of polycarbonate. Probes come with the temperature probe, 18" rod, and clamp to hold probe.
- Plage de température:Max. 500 °C
- Désignation:Hotplate 7×7", 120 V
- Maximum Capacity:10 L
- Top Plate Material:Ceramic, square