Microslide Viewer
Microslide viewer
Catalog # 470338-118
Supplier: SUPERTEK SCIENTIFIC LLC
Fournisseur #: T001
Jump to:
Product Details & Documents
The Microslide Viewer is specialty designed instrument for viewing Microslides®.
- Light weight and easy to use!
- No electricity or batteries required
Simply insert the slide and focus. The viewer brings 5X magnification allowing every image to be seen exactly as if observed through a microscope under ideal conditions. The white reflector provides sufficient light for a bright image in the normal light of a classroom. The viewer does not require electricity or batteries so it can also be used outdoors. 9″ high. For budding scientists of all ages!
Spécifications
- Désignation:Microslide viewer