Comparators for CHEMets® Test Kits
Supplier:   CHEMetrics
Comparators for CHEMets® Test Kits
Supplier:  CHEMetrics
Certificates
Comparators for CHEMets® Test Kits
Comparators for CHEMets® Test Kits
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
 : Store in the dark at room temperature.
Product Family Options
Informations sur le produit
  • Désignation
    Range
    Increments
    Availability
    Price
  • Ammonia High Range Flat Comparator
    1 - 10 ppm
    1, 2, 3, 4, 5, 6, 7, 8, 10 ppm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $69.35
    $69.35
    Each
       $69.35
    Supplier Number: C-1510
  • Filming Amines Round Comparator
    0 - 1 ppm
    0, 0.05, 0.10, 0.15, 0.25, 0.50, 0.75, 1.0 ppm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $43.75
    $43.75
    Each
       $43.75
    Supplier Number: C-1001
  • Low Range Ammonia Comparator
    0 - 1 ppm
    0, 0.1, 0.2, 0.3, 0.4, 0.6 0.8, 1.0 ppm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $47.83
    $47.83
    Each
       $47.83
    Supplier Number: C-1501
  • Phenol High Range Flat Comparator
    0 - 12 ppm
    0, 1, 2, 3, 4, 6, 8, 10, 12 ppm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $64.14
    $64.14
    Each
       $64.14
    Supplier Number: C-8012