The LyoPrime Luna™ Probe One-Step RT-qPCR Mix with UDG is offered in a lyophilized format, allowing for ambient/room temperature shipping and storage prior to use.
- Ambient/room temperature shipping and storage; two-year shelf life at room temperature
- Maximize your throughput by multiplexing up to five targets
- Reduce risk of carryover contamination with thermolabile UDG and dUTP included
- Unique passive reference dye offers compatibility across a variety of instrument platforms
- Contains non-fluorescent blue tracking dye for ease of pipetting
The mix contains all necessary components for one-step RT-qPCR except primers, probe(s), and RNA sample. It can be reconstituted by simply adding water to a 2 to 4X concentration, allowing for increased sample input. Performance in multiplexing applications has been optimized, with sensitive, linear detection achieved for up to five targets across a range of inputs.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.