ContactUsLinkComponent
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.45 µm
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.45 µm
Supplier:  Cytiva
Certificates
undefined
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.45 µm
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • CA28151-146
  • CA28151-142
  • CA28151-140
  • CA28151-138
  • CA28151-150
  • CA11012-908
  • CA11013-984
  • CA11013-584
  • CA11013-980
  • CA14230-030
  • CA97016-792
  • CA11008-530
  • CA30253-266
  • CA28297-734
  • CA28297-732
  • CA11013-524
  • CA11013-500
  • CA28159-525
  • CA11013-820
  • CA13500-162
  • CA13500-170
  • CA11008-580
  • CA89233-766
  • CA89233-768
  • CA89233-764
  • Weight:
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • 4.3 mg/cm² Minimum - 5.0 mg/cm² Maximum
  • Max. operating temperature:
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • 130 °C
  • Autoclavable:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Burst strength:
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • >0.68 bar
  • Porosity:
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • 74% minimum - 77% maximum
  • Protein:
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Medium protein binding
  • Solvent:
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
  • Medium solvent resistance
Specifications
Related Information
Frequently Bought Together