ContactUsLinkComponent
Accessories for VWR® Real Time Electrophoresis Systems
Catalog # 76518-302
Accessories for VWR® Real Time Electrophoresis Systems
Catalog # 76518-302
Supplier Number:  E1200-CS1-VWR
Specifications
  • Description:
    Extra casting set for 10.5×10 cm and 10.5×6 cm gels. Includes stand, trays, double-sided combs
  • Cat. no.:
    76518-302