SuperFlex™ PCR Plates, 96-Well, Low Profile, Simport
Supplier: Simport Scientific
Certificates
About this item
SuperFlex™ 'Break-Away' PCR plates allow for the most flexible, efficient and cost-effective option. Eliminate the use of scissors that cause perforated wells, damage sealing rings and could risk contamination issues.
- 0.2 ml low profile format
- For PCR and qPCR applications
- Compatible with all instruments that fit non-skirted, low profile plates
- Moulded from virgin polypropylene
- Sealable with the SIMPT321-96N PCR cap strips
The wells can be separated into 8 strips or higher saving time and money. You no longer have to throw away half empty plates after switching to the SuperFlex™.
The wells are individually flexible to have a perfect fit even when there is a difference in expansion coefficients. The SIMPT320-96W - White model improves signal reflection, reduces variation, and increases the sensitivity of qPCR assays.
Certifications: RNase, DNase, Human Genomic DNA free.