Translucent Snap Caps, Globe Scientific
À propos de cet article
These colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes and plastic culture tubes.
- Economical way to recap glass tubes, regardless of manufacturer
- Available in seven (7) translucent colors that correspond to Vacutainer® cap colors
- Caps can be written on with indelible ink for easy sample identification
- Prevents spills and evaporation by employing a liquid tight seal
These caps are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation and refrigeration procedures. They are available in bright translucent colors for sample identification purposes. The caps feature a single thumb tab for easy removal and close nesting in tube racks.
14 Options Available Below
Product Details & Documents
These colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes and plastic culture tubes.
- Economical way to recap glass tubes, regardless of manufacturer
- Available in seven (7) translucent colors that correspond to Vacutainer® cap colors
- Caps can be written on with indelible ink for easy sample identification
- Prevents spills and evaporation by employing a liquid tight seal
These caps are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation and refrigeration procedures. They are available in bright translucent colors for sample identification purposes. The caps feature a single thumb tab for easy removal and close nesting in tube racks.
Product Family Options
For 13 mm tubes
Informations sur le produit
- For Tube O.D.ColorAvailabilityPrice
- 13 mmBlueLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $62.68$62.68Prix promotionnel pour les nouveaux clientsPack of 1,000$62.68Supplier Number: 113143B
- 13 mmGrayLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $62.68$62.68Prix promotionnel pour les nouveaux clientsPack of 1,000$62.68Supplier Number: 113143A
- 13 mmGreenLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $62.68$62.68Prix promotionnel pour les nouveaux clientsPack of 1,000$62.68Supplier Number: 113143G
- 13 mmLavenderLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $62.68$62.68Prix promotionnel pour les nouveaux clientsPack of 1,000$62.68Supplier Number: 113143L
- 13 mmNaturalLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $62.68$62.68Prix promotionnel pour les nouveaux clientsPack of 1,000$62.68Supplier Number: 113143C
- 13 mmRedLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $62.68$62.68Prix promotionnel pour les nouveaux clientsPack of 1,000$62.68Supplier Number: 113143R
- 13 mmYellowLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $62.68$62.68Prix promotionnel pour les nouveaux clientsPack of 1,000$62.68Supplier Number: 113143Y
For 16 mm tubes
Informations sur le produit
- For Tube O.D.ColorAvailabilityPrice
- 16 mmBlueLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $66.88$66.88Prix promotionnel pour les nouveaux clientsPack of 1,000$66.88Supplier Number: 113144B
- 16 mmGrayLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $66.88$66.88Prix promotionnel pour les nouveaux clientsPack of 1,000$66.88Supplier Number: 113144A
- 16 mmGreenLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $66.98$66.98Prix promotionnel pour les nouveaux clientsPack of 1,000$66.98Supplier Number: 113144G
- 16 mmLavenderLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $68.74$68.74Prix promotionnel pour les nouveaux clientsPack of 1,000$68.74Supplier Number: 113144L
- 16 mmNaturalLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $68.74$68.74Prix promotionnel pour les nouveaux clientsPack of 1,000$68.74Supplier Number: 113144C
- 16 mmRedLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $67.41$67.41Prix promotionnel pour les nouveaux clientsPack of 1,000$67.41Supplier Number: 113144R
- 16 mmYellowLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $66.88$66.88Prix promotionnel pour les nouveaux clientsPack of 1,000$66.88Supplier Number: 113144Y