Metal to Glass Gasket, NW Flange, Ace Glass
NW (DN) flange metal to glass sealing gasket featuring a PTFE carrier and O-ring.
- High temperature metal to glass seal
- Made of PTFE
- Excellent for vaccum work or a maximum of 2 bar pressure
Included Capfe O-ring is suitable for use in temperatures from –60 to +204 °C. Viton O-Ring is suitable for use in temperatures from –26 to +240 °C.
Frequently Bought Together