BLACK-FIRE* Powder-Free Nitrile Exam Gloves, Halyard
Supplier: O&M HALYARD CANADA, INC CA
Certificates
About this item
BLACK-FIRE* Powder-Free Nitrile Exam Gloves combine high strength with tactile sensitivity, resulting in a glove that’s strong yet allows users to feel what they're doing.
- NFPA 1999-2013 Certified to protect against blood borne pathogens
- Superior tensile strength and elongation for increased protection and added comfort
- High-visibility orange inner layer shows through to warn the wearer of a rip or tear
- Exceeds current ASTM standards for critical defects, tensile strength, and puncture resistance
BLACK-FIRE* Nitrile Exam gloves are designed for superior protection and safety, whatever the task. These gloves are recommended for use in EMS, first responders, law enforcement, tattoo artists, dental, and healthcare industries.
Certifications: NFPA 1999 Certified, Compatible with the National Institute for Occupational Safety (NIOSH) recommendations and guidelines for handling fentanyl.
Specifications
- Catalog No:
- CA76540-320
- CA76540-318
- CA76540-316
- CA76540-314
- CA76540-312
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile