CELLvo™ Osteoarthritic Human Chondrocytes P0 (50-70y), StemBioSys
Supplier: StemBioSys
Certificates
About this item
CELLvo™ Ostoeoarthritic Human Chondrocytes (50-70y) are primary, uncultured cells isolated from recently-diseased cadaver cartilage from donors between 50 and 70 years old. Osteoarthritis status is determined by macroscopic inspection using a modified Outerbridge (1960) scale. Cells are isolated by enzymatic digestion prior to cryopreservation.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together