Volumetric Flask, Class A, Snap Cap, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
À propos de cet article
Durable, machine-blown body with heavy beaded tubing neck.
- Sharp graduation line and large white block lettering
A plastic snap cap is included.
Certifications: Complies with ASTM E288 Class A specifications; batch certified.
Frequently Bought Together