Order Entry
Canada
ContactUsLinkComponent
TeenPrep™ Metal Adapter, 12×15 ml, MP Biomedicals
TeenPrep™ Metal Adapter, 12×15 ml, MP Biomedicals
Catalog # CA76522-164
Supplier:  MP Biomedicals
undefined
TeenPrep™ Metal Adapter, 12×15 ml, MP Biomedicals
Catalog # CA76522-164
Supplier:  MP Biomedicals
Supplier Number:  116002546
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Capacity:
    12 x 15 ml tubes
  • Cat. no.:
    CA76522-164
  • For Use With:
    FastPrep-24, FastPrep-24 5G

Specifications

About this item

Metal adapter for FastPrep-24 instruments.

  • 12×15 ml tubes
  • Grind or homogenize difficult samples
  • Temperature resistance up to 450 °C

Due to its metal construction, the Metal TeenPrep™ Adapter can be run continuously on a FastPrep indefinitely, without the possibility of de-polymerization. This all-metal adapter can resist high temperatures up to 450 °C and can be sterilized by pyrolysis. Furthermore, the specially designed claws prevent tubes from auto-rotation during lysis, allowing continuous tracking of tube labels and/or bar codes.

The major use of this adapter is for lysing highly contagious samples and samples suspected of harboring prions. Due to its ability for high-temperature sterilization, this is the safest solution for CNS tissue lysis and other applications where prions or other disease-causing agents may be present.

The metal adapter is intended for difficult and long-lasting applications. The FastPrep bead beating system maximizes mechanical tissue homogenization for efficient isolation of high-quality DNA, RNA and proteins, without the need for chemicals or enzymes.

Accessories information: To secure this tube holder adapter on FastPrep 24 instruments, it is recommended to use the Ratchet Nut v3.0, blue (116005510), sold separately.
Caution: Unless specified otherwise, MP Biomedicals' products are for research or further manufacturing use only, not for direct human use.