Axygen® MaxyClear Snaplock Microcentrifuge Tube, Polypropylene, Spectrum, Non sterile, Corning
Fournisseur: Corning
Certificats
À propos de cet article
Axygen MaxyClear microcentrifuge tubes are available in a variety of colors. Axygen Maxymum Recovery® surface and sterile options are also available.
- 0,6, 1,5, and 2,0 ml clear microcentrifuge tubes are boil proof
- Colored tubes are not boil-proof
- Co-polymer tubes offer exceptional strength and chemical resistance
- Clear microcentrifuge tubes are boil-proof
- Maximum G-force is 14000 min⁻¹
- RNase-/DNase-free and nonpyrogenic
- Sterility Assurance Level (SAL) of 10⁻⁶
SnapLock microcentrifuge tubes have frosted cap surfaces and frosted side panels, ideal for labeling. Each cap features a piercing port in the center of the cap. Microcentrifuge tubes with no caps are available for some sizes.
Color - Assorted: Each pack contains a mixture of blue, green, violet, orange, red, and yellow.
Spécifications
- Catalog No:
- 80086-022
- 80086-024
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa