Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Fournisseur: Corning
Certificats
À propos de cet article
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
- Sera is aseptically collected and processed in a cGMP facility
- Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
- Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
- Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
- Premium FBS is sourced from C.A. herds
Spécifications
- Catalog No:
- CA76327-082
- CA76322-116
- CA76327-086
- Grade:
- Regular
- Regular
- Regular
- Origin:
- CA sourced
- CA sourced
- CA sourced
- Endotoxin:
- ≤20 EU/ml
- ≤20 EU/ml
- ≤20 EU/ml
- Hemoglobin:
- <30 mg/dl
- <30 mg/dl
- <30 mg/dl
- Storage temperature:
- −25...−15 °C
- −25...−15 °C
- −25...−15 °C