Quick-cfDNA/cfRNA Serum and Plasma Kit
Quick-cfDNA/RNA Serum and Plasma Kit, 50 Preps
Catalog # 76334-490
Supplier:   Zymo Research
 :  R1072
Quick-cfDNA/cfRNA Serum and Plasma Kit
Catalog # 76334-490
Supplier:  Zymo Research
CAS Number:  
Quick-cfDNA/cfRNA Serum and Plasma Kit
Quick-cfDNA/cfRNA Serum and Plasma Kit
Quick-cfDNA/RNA Serum and Plasma Kit, 50 Preps
CAS Number:  
Price
$1,630.44
Log in pour la disponibilité
Each
Product Details & Documents
The Quick-cfDNA/cfRNA Serum & Plasma Kit enables simple and efficient preparation of all circulating cell-free DNA and RNA (including protein-bound, exosomal, microRNA, and other small RNA) from serum, plasma, and other biological fluids, without using toxic phenol-chloroform or cumbersome protein precipitation and removal steps.
  • Simple spin-column based isolation from up to 3 mL of plasma, serum, or other biological liquids, no phenol/chloroform or precipitation
  • Efficient isolation of cell-free DNA and RNA. Recover up to 515 times more microRNA
  • Ready for Next-generation sequencing, RT-qPCR, nCounter, etc
The kit efficiently separates cell-free DNA and RNA, with option to co-elute. Cell-free nucleic acids scale linearly relative to the sample input volume up to 3 ml. Zymo-Spin™ Technology enables isolation of ultra-pure cell-free nucleic acids suitable for all subsequent analyses, such as RT-qPCR and NGS.
cfDNA and cfRNA can be eluted into 15 µl (to maximize total yield) or as little as 6 µl (for highly concentrated eluate) of DNase/RNase-Free Water.
Typically requires 30 minutes (cfDNA/cfRNA co-elution protocol) to 45 minutes (cfDNA/cfRNA separation protocol) hands-on time to process 10 samples. Sample digestion requires 2 hours.
High quality cell-free DNA (cfDNA) and cell-free RNA (cfRNA) are ready for all sensitive downstream applications such as RT-qPCR and Next-Generation Sequencing.
Up to 3 ml of plasma, serum, cerebrospinal fluids, amniotic fluids, urine, or other biological fluids.
 : Requires the following: Microcentrifuge, vortex, water bath or heating block, vacuum, and vacuum manifold.
 : Yields are sample dependent and may vary. Typical recovery ranges from 1 – 100 ng/ml of human plasma or serum for both cfDNA and cfRNA. Kit is optimized for recovery of total cfDNA and cfRNA, including small RNA and microRNA.
Spécifications
Spécifications
  • Désignation:
    Quick-cfDNA/RNA Serum and Plasma Kit
  • Taille:
    50 Preps
Spécifications