Reusable ESD StatMaster Labcoats, Long, ESD Knit Wrists, Uniform Technology
Supplier: Uniform Technology
Certificates
About this item
Reusable ESD labcoats are manufactured from 93% polyester/7% carbon.
- Lapel collar
- Snap front
- 2 waist pockets, 1 chest pocket
- ESD knit wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Specifications
- Catalog No:
- 76516-252
- 76516-250
- 76516-248
- 76516-246
- 76516-262
- 76516-260
- 76516-258
- 76516-256
- 76516-254
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Cleanroom
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon