Reusable ESD StatStar Labcoats, Long, ESD Knit Wrists, Uniform Technology
Supplier: Uniform Technology
Certificates
About this item
Reusable ESD labcoats are manufactured from 87% polyester/13% carbon.
- Lapel collar
- Snap front
- 2 waist pockets, 1 chest pocket
- ESD knit wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Specifications
- Catalog No:
- 76516-120
- 76516-136
- 76516-134
- 76516-132
- 76516-130
- 76516-128
- 76516-126
- 76516-124
- 76516-122
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon