Cole-Parmer® Essentials Plastic Jars, Wide-Mouth, HDPE, Antylia Scientific
Supplier: Antyila Scientific
Certificates
About this item
Get better access to samples with the wide-mouth design.
- Ideal for powder sampling
- 3.46" (88 mm) neck I.D.
Useful for storing dry chemicals. Jars are made of high-density polyethylene (HDPE) and include high-density polyethylene screw caps. Jars are not autoclavable.
Frequently Bought Together