Cole-Parmer® Essentials Carboy with Spigot, Rectangular, HDPE, Antylia Scientific
Don’t waste valuable bench space.
- Leakproof for safe transport
These leakproof HDPE rectangular carboys with spigot easily store and dispense sterile water and other solutions. Space-saving design requires less space than traditional round carboys. Features include built-in stainless steel handle and wide mouth opening for easy cleaning. Carboys are not autoclavable.
: USP Class VI certified
Frequently Bought Together