ContactUsLinkComponent
Reusable Altessa Grid Pull-Over Cleanroom Hoods with Face Mask, Uniform Technology
Reusable Altessa Grid Pull-Over Cleanroom Hoods with Face Mask, Uniform Technology
Supplier:  Uniform Technology
Certificates
Reusable Altessa Grid Pull-Over Cleanroom Hoods with Face Mask, Uniform Technology
Supplier:  Uniform Technology
Specifications
  • Catalog No:
  • 76515-942
  • 76515-940
  • 76515-938
  • 76515-936
  • 76515-934
  • 76515-932
  • 76515-930
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Fabric:
  • 99% polyester/1% carbon
  • 99% polyester/1% carbon
  • 99% polyester/1% carbon
  • 99% polyester/1% carbon
  • 99% polyester/1% carbon
  • 99% polyester/1% carbon
  • 99% polyester/1% carbon