Reusable Altessa Grid Cleanroom Boots with X-Stat ESD Sole, Uniform Technology
Cleanroom boots are manufactured from 99% polyester/1% carbon fabric.
- Color-coded by size webbing strap over toe and ankle
- Side release buckle
- X-Stat ESD molded sole
Suitable for ISO Class 5 or higher cleanrooms.
- Catalog No:
- 76515-852
- 76515-850
- 76515-848
- 76515-846
- 76515-844
- 76515-842
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Fabric:
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
Frequently Bought Together