Rubbermaid® Microfiber Floor Finishing System, Essendant
76500-254
- Capacity:3 gal.
- Dimensions:14.7D×16.2"H
- Material:Plastic/microfiber
- Cat. no.:76500-254
Minimize the potential for contamination by dedicating a color-coded system for floor finishing tasks. Kit includes everything needed to get started: finish pads, mop head frame, and bucket.
- Dedicated, color-coded floor finishing system
- Specially designed for smooth, even application
- Foam-lined microfiber pads
- Angled, washboard-surfaced bucket wall
- Lip-mounted handle stays
- One-step-change quick-connect frame
- Four 3" TPR swivel casters
When used in combination with quick-connect handles (sold separately), each piece works together to ensure smooth and even finish application. Foam-lined microfiber pads soak up more finish than standard mop heads while providing better liquid release control. Angled, washboard-surfaced bucket wall simplifies excess finish removal. Bucket's lip-mounted handle-sized notches keep mops upright and ready for use.