Argos Technologies® Reusable Inoculating Needles, Antylia Scientific
Fournisseur: Antyila Scientific
Certificats
À propos de cet article
Nichrome wire composition cools quickly and is less expensive than platinum.
- Select, inoculate, or transfer microbiological samples
Needles are made from high quality nichrome wire that cools rapidly after sterilization. Available with or without handle and straight or angled needle.
Frequently Bought Together