Reusable ESD StatMaster Labcoats, Long, Snap Wrists, Uniform Technology
Supplier: Uniform Technology
Certificates
About this item
Reusable ESD labcoats are manufactured from 93% polyester/7% carbon.
- Lapel collar
- Snap front
- 2 waist pockets, 1 chest pocket
- Snap wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Specifications
- Catalog No:
- 76516-186
- 76516-208
- 76516-184
- 76516-206
- 76516-182
- 76516-204
- 76516-180
- 76516-202
- 76516-178
- 76516-200
- 76516-198
- 76516-176
- 76516-174
- 76516-196
- 76516-194
- 76516-192
- 76516-190
- 76516-188
- Model:
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- BR5
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon
- 93% polyester/7% carbon