Reusable Altessa Grid Snap-Front Cleanroom Hoods, Uniform Technology
Supplier: Uniform Technology
Certificates
About this item
Cleanroom hoods are manufactured from 99% polyester/1% carbon fabric.
- Open face design
- Snaps under chin
- Vertical and horizontal snaps on back
- Snaps at temples
Suitable for ISO Class 5 or higher cleanrooms.
Specifications
- Catalog No:
- 76515-818
- 76515-840
- 76515-838
- 76515-834
- 76515-832
- 76515-830
- 76515-828
- 76515-826
- 76515-824
- 76515-822
- 76515-820
- 76515-816
- 76515-814
- 76515-836
- Model:
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- CHM-74
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon