2010 Geno/Grinder® Homogenizer Adjustable Clamp, SPEX SamplePrep
Catalog # 76533-110
Supplier: SPEX
Specifications
- Description:Geno/Grinder® replacement adjustable clamp assembly
- Cat. no.:76533-110
- For Use With:HG-600 homogenizer
- Details:Clamp provides 1.25" (3.2 cm) vertical movement
Specifications
About this item
Adjusts to fit various sizes of titer plates and vials.
- Accommodates up to six deep-well titer plates or up to sixteen 50 ml tubes
- Stack titer plates in the clamp increasing throughput
- Release mechanism makes it easy to slide clamp up and down the shaft
- Adjustable Clamp Assembly for new model 2010 Geno/Grinder
- Clamp provides 1.25" (3.2 cm) vertical movement
This front loading clamp is included as standard in all new 2010 Geno/Grinders but can be purchased separately as a retro-fit.