Vikan® One-Piece Shovel, Remco
Supplier: Remco Products
Jump to:
À propos de cet article
One-piece polypropylene shovels are tough, lightweight and hygienic.
- Popular for meat, dairy, seafood and poultry processing
- Also suitable for commercial fishing and other food handling applications
- Available in 10 or 14" blade widths
- Polypropylene construction
Molded from FDA-compliant polypropylene, it is a workhorse for moving bulk materials and an integral part of any professional cross–contamination containment, food safety or HACCP program.
6 Options Available Below
Product Details & Documents
One-piece polypropylene shovels are tough, lightweight and hygienic.
- Popular for meat, dairy, seafood and poultry processing
- Also suitable for commercial fishing and other food handling applications
- Available in 10 or 14" blade widths
- Polypropylene construction
Molded from FDA-compliant polypropylene, it is a workhorse for moving bulk materials and an integral part of any professional cross–contamination containment, food safety or HACCP program.
Product Family Options
Informations sur le produit
- ColorDimensionsAvailabilityPrice
- Supplier Number: 698166
- Supplier Number: 698188
- Supplier Number: 698177
- Supplier Number: 698266
- Supplier Number: 698288
- Supplier Number: 698277