Vikan® High Temperature Scrapers, Remco
Supplier: Remco Products
Certificates
About this item
These high temperature large hand scrapers are great for scraping hot cooking surfaces.
- For use in temperatures up to 410 °F
- Nylon/glass fiber construction
- Seamless design
- Length: 9.38"
The scraper has no seams or cracks, which helps prevent bacterial growth.
Frequently Bought Together